Skip to Content
Merck
All Photos(1)

Key Documents

407272

Sigma-Aldrich

Integrin αvβ3 Antagonist, P11

The Integrin αvβ3 Antagonist, P11 controls the biological activity of Integrin αvβ3.

Synonym(s):

Integrin αvβ3 Antagonist, P11, HSDVHK-NH2, HSDVHK

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€162.00

€162.00


Estimated to ship on25 May 2025



Select a Size

Change View
5 MG
€162.00

About This Item

Empirical Formula (Hill Notation):
C30H51N12O93 · 3C2HF3O2
Molecular Weight:
2409.82
UNSPSC Code:
12352200
NACRES:
NA.77

€162.00


Estimated to ship on25 May 2025


Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

water: 100 mg/mL

shipped in

wet ice

storage temp.

−20°C

General description

A cell-permeable, hexapeptide containing a novel integrin-binding motif, SDV (a Type I PDZ-binding motif) that is shown to compete with vitronectin for the RGD-binding site of integrin αvβ3 (IC50 = 1.74 pg/ml), in a chip-based assay. It is rapidly internalized into HUVECs, but not in NIH 3T3 cells, at a culture temperature of 37 °C, a process which is mediated by caveolin and clathrin. At 10 µg/ml, this compound is shown to significantly inhibit the phosphorylation of ERK1/2 and MEK in HUVECs. Furthermore, it suppresses bFGF-induced proliferation of HUVECs, dose-dependently, and displays up-regulation of p53 and induction of apoptosis via activation of caspases-3, -8 and -9 at 50 µg/ml.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Physical form

Supplied as a trifluoracetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Bang, J., et al. 2011. Mol Cell Proteomics10, M110.
Choi, Y., et al. 2010. Proteomics10, 72.
Lee, Y., et al. 2004. J. Biomol. Screen.9, 687.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eunji Jo et al.
Biotechnology and bioengineering, 115(6), 1437-1449 (2018-02-21)
The endocytosis-mediating performances of two types of peptide ligands, cell receptor binding peptide (CRBP) and cell membrane penetrating peptide (CMPP), were analyzed and compared using a common carrier of peptide ligands-human ferritin heavy chain (hFTH) nanoparticle. Twenty-four copies of a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service