Skip to Content
Merck
All Photos(1)

Key Documents

407137

Sigma-Aldrich

D-myo-Inositol 1,4,5-Trisphosphate, trisodium salt

≥95% (TLC), solid, second messenger involved in Ca2+ mobilization, Calbiochem®

Synonym(s):

D-myo-Inositol 1,4,5-Trisphosphate, Trisodium Salt, D-Ins(1,4,5)P₃, 3Na

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€214.00

€214.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
1 MG
€214.00

About This Item

Empirical Formula (Hill Notation):
C6H12O15P3 · 3Na
CAS Number:
Molecular Weight:
486.04
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.77

€214.00


Estimated to ship on30 May 2025


Request a Bulk Order

Product Name

D-myo-Inositol 1,4,5-Trisphosphate, Trisodium Salt, Sodium salt of D-Ins(1,4,5)P₃ that has similar biological properties as the lithium salt.

Quality Level

Assay

≥95% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

water: 10 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[P](=O)([O-])([O-])O[C@H]1[C@@H]([C@H]([C@H]([C@H]([C@@H]1O)O[P](=O)([O-])[O-])O)O)O[P](=O)([O-])[O-]

InChI

1S/C6H15O15P3.6Na/c7-1-2(8)5(20-23(13,14)15)6(21-24(16,17)18)3(9)4(1)19-22(10,11)12;;;;;;/h1-9H,(H2,10,11,12)(H2,13,14,15)(H2,16,17,18);;;;;;/q;6*+1/p-6/t1-,2+,3+,4-,5-,6-;;;;;;/m1....../s1

InChI key

MLMBGBMAFNSCOW-PPKFVSOLSA-H

Related Categories

General description

Sodium salt of D-Ins(1,4,5)P3 that has similar biological properties as the lithium salt (Cat. No. 407123).
Sodium salt of D-Ins(1,4,5)P3 that has similar biological properties as the lithium salt (Cat. No. 407123).

Biochem/physiol Actions

Cell permeable: no
Primary Target
Mobilization of Ca2+ from intracellular stores.
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ka-Chun Mok et al.
Clinical and translational medicine, 11(11), e578-e578 (2021-11-30)
Hyperpolarization-activated cyclic nucleotide-gated (HCN) channels are members of the voltage-gated cation channel family known to be expressed in the heart and central nervous system. Ivabradine, a small molecule HCN channel-blocker, is FDA-approved for clinical use as a heart rate-reducing agent.
Jone Paesmans et al.
eLife, 9 (2020-12-23)
Synaptojanin1 (Synj1) is a phosphoinositide phosphatase, important in clathrin uncoating during endocytosis of presynaptic vesicles. It was identified as a potential drug target for Alzheimer's disease, Down syndrome, and TBC1D24-associated epilepsy, while also loss-of-function mutations in Synj1 are associated with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service