Skip to Content
Merck
All Photos(1)

Key Documents

401253

Sigma-Aldrich

Goat Anti-Mouse IgG, H&L Chain Specific Peroxidase Conjugate

liquid, Calbiochem®

Synonym(s):

Anti-Mouse IgG Peroxidase

Sign Into View Organizational & Contract Pricing

Select a Size

2 ML
€335.00

€335.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
2 ML
€335.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

€335.00


Estimated to ship on26 May 2025


Request a Bulk Order

biological source

goat

Quality Level

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

liquid

contains

0.02% thimerosal as preservative

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

isotype

IgG

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

General description

Adsorbed against bovine, horse, and human serum proteins.
Immunoaffinity purified goat polyclonal antibody conjugated to horseradish peroxidase. Adsorbed against bovine, horse, and human serum proteins. Recognizes mouse IgG, heavy and light chains.
This Goat Anti-Mouse IgG, H&L Chain Specific Peroxidase Conjugate is validated for use in Enzyme Immunoassay, Immunoelectrophoresis for the detection of Mouse IgG, H&L Chain Specific.

Immunogen

Mouse

Application

Enzyme Immunoassay (1:10,000)

Immunoelectrophoresis (see comments)

Warning

Toxicity: Standard Handling (A)

Physical form

In 10 mM PBS, 0.5 mM EDTA, 10% mannose, 2% sucrose, 1% BSA, pH 7.6.

Other Notes

Monospecific for mouse IgG, heavy and light chains, as determined by immunoelectrophoresis against normal mouse serum. Cross-reactivity to normal bovine, horse and human sera: <1% by direct solid phase immunoassay. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

To comply with ban of sale of mercury-added products required by The Interstate Mercury Education and Reduction Clearinghouse (IMERC), this product is prohibited to be sold in the following US states: Rhode Island and Connecticut.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Frances E Carr et al.
Endocrinology, 157(8), 3278-3292 (2016-06-03)
Dysregulation of the thyroid hormone receptor (TR)β is common in human cancers. Restoration of functional TRβ delays tumor progression in models of thyroid and breast cancers implicating TRβ as a tumor suppressor. Conversely, aberrant expression of the runt-related transcription factor
Margarida Barroso et al.
The Journal of biological chemistry, 290(45), 27101-27112 (2015-09-25)
Antigen processing and MHC class II-restricted antigen presentation by antigen-presenting cells such as dendritic cells and B cells allows the activation of naïve CD4+ T cells and cognate interactions between B cells and effector CD4+ T cells, respectively. B cells
Kyoko Ito et al.
Cell reports, 28(10), 2480-2490 (2019-09-05)
The Ten-eleven translocation (TET) enzymes regulate gene expression by promoting DNA demethylation and partnering with chromatin modifiers. TET2, a member of this family, is frequently mutated in hematological disorders. The contributions of TET2 in hematopoiesis have been attributed to its
Natalia Di Pietro et al.
Molecular and cellular biochemistry, 417(1-2), 155-167 (2016-05-22)
Red blood cells (RBCs) enzymatically produce nitric oxide (NO) by a functional RBC-nitric oxide synthase (RBC-NOS). NO is a vascular key regulatory molecule. In RBCs its generation is complex and influenced by several factors, including insulin, acetylcholine, and calcium. NO
Pradeep K Singh et al.
PloS one, 10(3), e0120346-e0120346 (2015-03-25)
Conversion of amyloid fibrils by many peptides/proteins involves cytotoxic helix-rich oligomers. However, their toxicity and biophysical studies remain largely unknown due to their highly dynamic nature. To address this, we chose two helical peptides (melittin, Mel and pancreatic polypeptide, PP)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service