Skip to Content
Merck
All Photos(1)

Key Documents

401135

Sigma-Aldrich

Goat Anti-Human IgA, α-Chain Specific Peroxidase Conjugate

liquid, Calbiochem®

Sign Into View Organizational & Contract Pricing

Select a Size

2 ML
€264.00

€264.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
2 ML
€264.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.77

€264.00


Estimated to ship on24 May 2025


Request a Bulk Order

biological source

goat

Quality Level

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

liquid

contains

0.02% thimerosal as preservative

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

isotype

IgG

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

General description

Immunoaffinity purified goat polyclonal antibody conjugated to horseradish peroxidase. Recognizes human IgA, α-chain.
This Goat Anti-Human IgA, α-Chain Specific Peroxidase Conjugate is validated for use in ELISA for the detection of Human IgA, α-Chain Specific.

Immunogen

Human

Application

ELISA (1:10,000 for direct ELISA)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 10 mM PBS, 500 µM EDTA, 10% mannose, 2% sucrose, 1% BSA, pH 8.0.

Other Notes

Monospecific for human immunoglobulin A, α-chain against normal human serum as measured by ELISA. Each lot of antibody is tested by ELISA, but is applicable to any immunoassay requiring a peroxidase-conjugated secondary antibody. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

To comply with ban of sale of mercury-added products required by The Interstate Mercury Education and Reduction Clearinghouse (IMERC), this product is prohibited to be sold in the following US states: Rhode Island and Connecticut.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoqi Yang et al.
STAR protocols, 3(1), 101203-101203 (2022-03-15)
Antibodies in milk obtained from those previously SARS-CoV-2-infected or vaccinated against COVID-19 may provide passive immunity to the breastfed infant. Few assays have been established to measure antibodies in human milk, despite the public health importance of this topic. In

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service