Skip to Content
Merck
All Photos(1)

Key Documents

400088

Sigma-Aldrich

HIF-1α Translation Inhibitor, KC7F2

The HIF-1α Translation Inhibitor, KC7F2, also referenced under CAS 927822-86-4, controls the biological activity of HIF-1α. This small molecule/inhibitor is primarily used for Cell Structure applications.

Synonym(s):

HIF-1α Translation Inhibitor, KC7F2, Hypoxia-Inducible Factor-1α Translation Inhibitor, KC7F2, N,Nʹ-(2,5-Dichlorosulfonyl)cystamine, HIF Inhibitor III

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€188.00

€188.00


Estimated to ship on28 May 2025



Select a Size

Change View
25 MG
€188.00

About This Item

Empirical Formula (Hill Notation):
C16H16Cl4N2O4S4
CAS Number:
Molecular Weight:
570.38
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€188.00


Estimated to ship on28 May 2025


Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

beige to tan

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

[S](=O)(=O)(NCCSSCCN[S](=O)(=O)c2c(ccc(c2)Cl)Cl)c1c(ccc(c1)Cl)Cl

InChI

1S/C16H16Cl4N2O4S4/c17-11-1-3-13(19)15(9-11)29(23,24)21-5-7-27-28-8-6-22-30(25,26)16-10-12(18)2-4-14(16)20/h1-4,9-10,21-22H,5-8H2

InChI key

REQLACDIZMLXIC-UHFFFAOYSA-N

General description

A cell-permeable symmetrical cystamine compound that selectively suppresses cellular HIF-1α, but not HIF-1β, protein synthesis without affecting the transcription of HIF-1α mRNA or the stability of HIF-1α protein. Reduction of cellular HIF-1α by KC7F2 treatment (IC50<20 µM in LN229 cultures under hypoxia condition) is shown to result in decreased transcriptions of HIF-1α-dependent genes, such as CA IX, endothelin 1, enolase 1, and MMP-2. Inhibition of 4EBP1 and S6K phosphorylation, important event for the initiation of protein translation, is also observed in KC7F2 and U87MGD cultures following KC7F2 treatment under hypoxia condition, which may account for, at least in part, the downregulated HIF-1α synthesis.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Narita, T., et al. 2009. Clin. Cancer Res.15, 6128.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service