Skip to Content
Merck
All Photos(1)

Key Documents

390900

Sigma-Aldrich

4-Hydroxyphenylretinamide

A synthetic amide of all-trans retinoic acid (RA) that displays reduced toxicity relative to RA while maintaining significant biological activity.

Synonym(s):

4-Hydroxyphenylretinamide, Fenretinide, 4HPR, N-(4-Hydroxyphenyl)-all- trans Retinamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€205.00

€205.00


Estimated to ship on21 May 2025



Select a Size

Change View
5 MG
€205.00

About This Item

Empirical Formula (Hill Notation):
C26H33NO2
CAS Number:
Molecular Weight:
391.55
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€205.00


Estimated to ship on21 May 2025


Quality Level

description

Merck USA index - 14, 3998

Assay

≥97% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

orange

solubility

DMSO: 25 mg/mL
acetone: soluble
ethanol: soluble

shipped in

ambient

storage temp.

−20°C

SMILES string

N(c2ccc(cc2)O)C(=O)\C=C(/C)\C=C\C=C(/C)\C=C\C1=C(CCCC1(C)C)C

InChI

1S/C26H33NO2/c1-19(11-16-24-21(3)10-7-17-26(24,4)5)8-6-9-20(2)18-25(29)27-22-12-14-23(28)15-13-22/h6,8-9,11-16,18,28H,7,10,17H2,1-5H3,(H,27,29)/b9-6+,16-11+,19-8+,20-18+

InChI key

AKJHMTWEGVYYSE-FXILSDISSA-N

General description

A synthetic amide of all-trans retinoic acid (RA) that displays reduced toxicity relative to RA while maintaining significant biological activity. Active in the prevention and treatment of a variety of neoplasms in animals. Inhibits proliferation in cell lines of human breast cancer, prostate adenocarcinoma, and ovarian carcinoma. Inhibits angiogenesis, as well as endothelial cell motility and tubule formation. Induces apoptosis in malignant hemopoietic cell lines.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Active in the prevention and treatment of a variety of neoplasms in animals.
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful & Carcinogenic / Teratogenic (E)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Formelli, F., et al. 1996. FASEB J. 10, 1014.
Naik, H.R., et al. 1995. Adv. Pharmacol. 33, 315.
Delia, D., et al. 1993. Cancer Res. 53, 6036.
Pienta, K.J., et al. 1993. Cancer Res. 53, 224.
Grubbs, C.J., et al. 1990. Anticancer Res. 10, 661.
McCarthy, D.J., et al. 1987. Cancer Res.47, 5014.
Moon, R.C., et al. 1979. Cancer Res.39, 1339.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service