Skip to Content
Merck
All Photos(1)

Key Documents

385908

Millipore

Hyaluronic Acid, Sodium Salt, Streptococcus pyrogenes

Natural high-viscosity mucopolysaccharide with alternating β1,3-glucuronidic and β1,4-glucosaminidic bonds. Principal glycosaminoglycan in connective tissue fluids.

Synonym(s):

Hyaluronic Acid, Sodium Salt, Streptococcus pyrogenes

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
Inquire


Estimated to ship on03 April 2025


Request a Bulk Order

Select a Size

Change View
500 MG
Inquire

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.79


Estimated to ship on03 April 2025


Request a Bulk Order

Quality Level

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

impurities

≤0.070 EU/mg endotoxins (EU/mg NaHy)
≤0.1% protein

solubility

water: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[Na+].N([C@H]1[C@@H](O[C@@H]([C@H]([C@@H]1O[C@@H]4O[C@@H]([C@H]([C@@H]([C@H]4O)O)O)C(=O)O)O)CO)O[C@@H]2[C@H](O[C@H]([C@@H]([C@H]2O)O)O[C@@H]3[C@H]([C@@H](O[C@@H]([C@H]3O)CO)O)NC(=O)C)C(=O)O)C(=O)C

InChI

1S/C28H44N2O23.Na/c1-5(33)29-9-18(11(35)7(3-31)47-25(9)46)49-28-17(41)15(39)20(22(53-28)24(44)45)51-26-10(30-6(2)34)19(12(36)8(4-32)48-26)50-27-16(40)13(37)14(38)21(52-27)23(42)43;/h7-22,25-28,31-32,35-41,46H,3-4H2,1-2H3,(H,29,33)(H,30,34)(H,42,43)(H,44,45);/q;+1/t7-,8-,9-,10-,11-,12-,13+,14+,15-,16-,17-,18-,19-,20+,21+,22+,25-,26+,27-,28-;/m1./s1

InChI key

YWIVKILSMZOHHF-QJZPQSOGSA-N

General description

Natural high-viscosity mucopolysaccharide with alternating β1,3-glucuronidic and β1,4-glucosaminidic bonds. Principal glycosaminoglycan in connective tissue fluids.
Natural high-viscosity mucopolysaccharide with alternating β1,3-glucuronidic and β1,4-glucosaminidic bonds. Principal glycosaminoglycan in connective tissue fluids. The molecular weight of Hyaluronic Acid, Sodium Salt, is in the range of 5.0 x 105 - 1.2 x 106, as calculated from its limiting viscosity.

Application


  • Hyaluronic Acid as a LYVE-1 Receptor Ligand in the Lymphatic System of Healthy Human Skin.: This study investigates the role of hyaluronic acid as a ligand for the LYVE-1 receptor in the lymphatic system, offering insights into its physiological roles and potential therapeutic applications, particularly in treating lymphatic-related diseases (Michurina et al., 2024).

  • M2 Macrophage-Derived Extracellular Vesicles Encapsulated in Hyaluronic Acid Alleviate Osteoarthritis by Modulating Macrophage Polarization.: This research explores the therapeutic potential of M2 macrophage-derived extracellular vesicles encapsulated in hyaluronic acid for osteoarthritis treatment. It highlights a novel anti-inflammatory mechanism that could be significant for biochemists studying cellular therapies (Yuan et al., 2024).

  • Insights into Translational and Biomedical Applications of Hydrogels as Versatile Drug Delivery Systems.: This review covers the use of hyaluronic acid in hydrogel drug delivery systems, emphasizing its utility in various biomedical applications. It provides a detailed analysis of the material′s properties, applications, and future potential in drug delivery and tissue engineering (Kohar et al., 2024).

  • Utilizing Ni(II) complex for metal drug-gel particles in cervical cancer treatment and designing novel drugs through machine learning methods.: Although primarily focused on Ni(II) complex, this study discusses the integration of metal drug-gel particles with hyaluronic acid, offering insights into its potential as a carrier for targeted therapy in cervical cancer (Jiang et al., 2024).

Warning

Toxicity: Standard Handling (A)

Preparation Note

Heating slowly to 50°C may be required for complete solubilization.

Reconstitution

Following reconstitution, refrigerate (4°C). Stock solutions are stable for up to 3 months at 4°C in the absence of contamination.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service