Skip to Content
Merck

Skip To

382425

Trequinsin, Hydrochloride

Extremely potent and cell-permeable inhibitor of cGMP-inhibited phosphodiesterase (IC50 = 300 pM) and platelet aggregation in vitro.

Synonym(s):

Trequinsin, Hydrochloride, 9,10-Dimethoxy-2-mesitylimino-3-methyl-2,3,6,7-tetrahydro-4H-pyrimido-(6,1-a)-isoquinolin-4-one, HCl, HL 725

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 mg
Check Cart for Availability
€ 327,00

About This Item

Empirical Formula (Hill Notation):
C24H27N3O3 · xHCl
CAS Number:
Molecular Weight:
405.49 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
solid
Storage condition:
OK to freeze, desiccated

€ 327,00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

description

RTECS - UW7727000

assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, desiccated

color

pale yellow

solubility

water: 500 μM

shipped in

ambient

storage temp.

10-30°C

SMILES string

Cl.[n]21c(c[c]([n]([c]2=O)C)=Nc4c(cc(cc4C)C)C)c3c(cc(c(c3)OC)OC)CC1

InChI

1S/C24H27N3O3.ClH/c1-14-9-15(2)23(16(3)10-14)25-22-13-19-18-12-21(30-6)20(29-5)11-17(18)7-8-27(19)24(28)26(22)4;/h9-13H,7-8H2,1-6H3;1H

InChI key

DTCZZBVPTHVXFA-UHFFFAOYSA-N

General description

Extremely potent and cell-permeable inhibitor of cGMP-inhibited phosphodiesterase (IC50 = 300 pM) and platelet aggregation in vitro. Potentiates adenosine-stimulated cAMP accumulation.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
cGMP-inhibited phosphodiesterase
Product does not compete with ATP.
Reversible: no
Target IC50: 300 pM against cGMP-inhibited phosphodiesterase

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Spina, D., et al. 1998. Life Sci. 62, 953.
Pillai, R., et al. 1994. J. Biol. Chem.269, 30676.
Tang, K.M., et al. 1994. Eur. J. Pharmacol.268, 105.
Whalin, M.E., et al. 1991. Mol. Pharmacol. 39, 711.
Beavo, J.A., and Reifsnyder, D.H. 1990. Trends Pharmacol. Sci.11, 150.
DeClerck, F., et al. 1990. Blood Coag. Fibrinol. 1, 247.
Konkle, B.A., et al. 1990. J. Biol. Chem. 265, 21867.
Ruppert, D., and Weithmann, K.U. 1982. Life Sci.31, 2037.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
551850215921T2057
form

solid

form

solid

form

crystalline solid

form

powder

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥99% (HPLC)

assay

≥98%

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

-

storage temp.

10-30°C

storage temp.

10-30°C

storage temp.

10-30°C

storage temp.

-

solubility

water: 500 μM

solubility

water: soluble

solubility

methanol: 10 mg/mL, water: 50 mg/mL

solubility

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters