Skip to Content
Merck
All Photos(1)

Key Documents

382172

Sigma-Aldrich

Histone Deacetylase Inhibitor VI, HNHA

The Histone Deacetylase Inhibitor VI, HNHA, also referenced under CAS 926908-04-5, controls the biological activity of Histone Deacetylase.

Synonym(s):

Histone Deacetylase Inhibitor VI, HNHA, N-Hydroxy-7-(2-naphthylthio)heptanomide

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€190.00

€190.00


Estimated to ship on28 May 2025



Select a Size

Change View
10 MG
€190.00

About This Item

Empirical Formula (Hill Notation):
C17H21NO2S
CAS Number:
Molecular Weight:
303.42
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.54

€190.00


Estimated to ship on28 May 2025


Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

methanol: 10 mg/mL
DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

S(CCCCCCC(=O)NO)c1cc2c(cc1)cccc2

InChI

1S/C17H21NO2S/c19-17(18-20)9-3-1-2-6-12-21-16-11-10-14-7-4-5-8-15(14)13-16/h4-5,7-8,10-11,13,20H,1-3,6,9,12H2,(H,18,19)

InChI key

KPNNXHVGOKRBEF-UHFFFAOYSA-N

General description

A cell-permeable hydroxyamide-based SAHA analog that potently inhibits histone deacetylase (HDAC) activity (IC50 = 100 nM). Shown to induce histone acetylation and p21 transcription with concomitent inhibition of cell cycle progression in human fibrosarcoma HT1080 cells. Reported to be more potent than SAHA in inhibiting tumor growth in a murine xenograph model in vivo.

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Kim, D.H., et al. 2007. Biochem. Biophys. Res. Commun.356, 233.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dong Hoon Kim et al.
Biochemical and biophysical research communications, 356(1), 233-238 (2007-03-14)
Histone deacetylase (HDAC), a key enzyme in gene expression and carcinogenesis, is considered an attractive target molecule for cancer therapy. Here, we report a new synthetic small molecule, N-hydroxy-7-(2-naphthylthio) heptanomide (HNHA), as a HDAC inhibitor with anti-tumor activity both in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service