Skip to Content
Merck
All Photos(1)

Key Documents

382111

Sigma-Aldrich

Histone Acetyltransferase Inhibitor IV, CPTH2

Synonym(s):

Histone Acetyltransferase Inhibitor IV, CPTH2, HAT Inhibitor IV, Cyclopentylidene-(4-(4ʹ-chlorophenyl)thiazol-2-yl)hydrazone, GCN5 Inhibitor II, N-acetyltransferase 10 Inhibitor I, NAT10 Inhibitor I

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€123.00

€123.00


Estimated to ship on28 May 2025



Select a Size

Change View
10 MG
€123.00

About This Item

Empirical Formula (Hill Notation):
C14H14ClN3S
CAS Number:
Molecular Weight:
291.80
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.54

€123.00


Estimated to ship on28 May 2025


Assay

≥95% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

brown

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C14H14ClN3S/c15-11-7-5-10(6-8-11)13-9-19-14(16-13)18-17-12-3-1-2-4-12/h5-9H,1-4H2,(H,16,18)

InChI key

YYTHPXHGWSAKIZ-UHFFFAOYSA-N

General description

A cell-permeable thiazolyl-hydrazone compound that inhibits the acetylation of histone H3 by recombinant human Gcn5 in a substrate-competive manner, while exhibiting little activity against rhGcn5 autoacetylation. Although functional knockout of Gcn5 in Saccharomyces cerevisiae by genetic deletion or by CPTH2 treatment is not detrimental to the survival of the budding yeast, CPTH2 treatment is shown to inhibit the growth of yeast strain genetically deleted of Gcn5, but not strains deleted of Elp3 or Sas2.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Larrieu, D., et al. 2014. Science, 344, 527.
Chimenti, F., et al. 2009. J. Med. Chem.52, 530.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joshy George et al.
Cancer research communications, 2(6), 402-416 (2023-01-24)
The emergence of treatment resistance significantly reduces the clinical utility of many effective targeted therapies. Although both genetic and epigenetic mechanisms of drug resistance have been reported, whether these mechanisms are stochastically selected in individual tumors or governed by a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service