Skip to Content
Merck
All Photos(1)

Key Documents

3810CBC

Millipore

L-Histidine

free base

Synonym(s):

L-Histidine, (S)-2-Amino-3-(4-imidazolyl)propionic acid, NSC 137773

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9N3O2
CAS Number:
Molecular Weight:
155.15
Beilstein:
84088
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.26
Pricing and availability is not currently available.

Quality Level

mp

282 °C (dec.) (lit.)

SMILES string

N[C@@H](Cc1c[nH]cn1)C(O)=O

InChI

1S/C6H9N3O2/c7-5(6(10)11)1-4-2-8-3-9-4/h2-3,5H,1,7H2,(H,8,9)(H,10,11)/t5-/m0/s1

InChI key

HNDVDQJCIGZPNO-YFKPBYRVSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Precursor of histamine by action of histidine decarboxylase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service