Skip to Content
Merck
All Photos(1)

Key Documents

374807

Sigma-Aldrich

Hemocyanin, Keyhole Limpet, Megathura crenulata, BES/Mg2+ Buffer

Hemocyanin, Keyhole Limpet, Megathura crenulata, BES/Mg2+ buffer, CAS 9013-72-3, is a large, multi-subunit, O2-carrying, metalloprotein that is used in the production of antibodies.

Synonym(s):

Hemocyanin, Keyhole Limpet, Megathura crenulata, BES/Mg2+ Buffer, KLH

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€474.00

€474.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
250 MG
€474.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.41

€474.00


Estimated to ship on28 May 2025


Request a Bulk Order

Quality Level

Assay

≥90% (size exclusion chromatography)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

color

clear blue-gray

shipped in

wet ice

storage temp.

2-8°C

General description

Keyhole limpet hemocyanin (KLH) is a large, multi-subunit, O2-carrying, metalloprotein. It is isolated from a marine snail species called Megathura crenulate.[1]

Biochem/physiol Actions

Keyhole limpet hemocyanin (KLH) exhibits immunostimulatory properties and stimulates both cell-mediated and humoral responses in animals and humans.[1] It serves as a vaccine adjuvant[1][2] and facilitates protein conjugation. In addition, KLH acts as a carrier protein to produce antibodies as well as haptens such as peptides and oligosaccharides. It is also used in the treatment of bladder cancer.[2]

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 10 mM MgSO₄, 5 mM BES buffer, 0.9% NaCl, pH 6.5. Sterile-filtered.

Preparation Note

KLH is an extremely large protein complex that has a tendency to aggregate and precipitate from solutions. Use a buffer when diluting KLH and maintain the magnesium concentration at ≥10 mM. Store KLH solutions at 4°C. DO NOT freeze, vortex or agitate to dissolve KLH. Freezing or agitating KLH will cause protein aggregation and precipitation.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evaluation of human antibody responses to keyhole limpet hemocyanin on a carbohydrate microarray
Oyelaran O and Gildersleeve JC, et al.
4th International Conference on Biomedical Engineering in Vietnam, 285-294 (2010)
Oyindasola Oyelaran et al.
Proteomics. Clinical applications, 4(3), 285-294 (2010-12-08)
Keyhole limpet hemocyanin (KLH) is used as a vaccine adjuvant, as a carrier protein for small haptens, and as a treatment for bladder cancer. Immunization with KLH produces antibodies to tumor-associated carbohydrate antigens (TACAs) in animals, and these antibodies have
Erik H J G Aarntzen et al.
Cancer immunology, immunotherapy : CII, 61(11), 2003-2011 (2012-04-25)
Keyhole limpet hemocyanin (KLH) attracts biomedical interest because of its remarkable immunostimulatory properties. Currently, KLH is used as vaccine adjuvant, carrier protein for haptens and as local treatment for bladder cancer. Since a quantitative human anti-KLH assay is lacking, it

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service