Skip to Content
Merck
All Photos(1)

Key Documents

3730-OP

Millipore

Dextran Sulfate

OmniPur® Grade, Dextran Sulfate, 50% Solution Autoclaved.

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
Inquire


Estimated to ship on27 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
Inquire

About This Item

UNSPSC Code:
12352201
NACRES:
NA.28


Estimated to ship on27 May 2025


Request a Bulk Order

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

haze-free tan to yellow

shipped in

ambient

storage temp.

10-30°C

General description

Specifications:
DNase: None detected
Protease: None detected
RNase: None detected




Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. A quick question, Is the 50% the weight percentage or volume percentage?

    1 answer
    1. The concentration is expressed as weight percentage.

      Helpful?

  2. What is the molecular weight of this sulfated dextran?

    1 answer
    1. The molecular weight for product 3730-OP is 500,000 Da.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service