Skip to Content
Merck
All Photos(1)

Key Documents

371209

Sigma-Aldrich

Dkk-2, Mouse, Recombinant

Dkk-2, Mouse, Recombinant, consists of amino acids 26-259 and expressed in S. frugiperda 21 cells with a C-terminal His•Tag sequence.

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€539.00

€539.00


Estimated to ship on30 April 2025



Select a Size

Change View
10 μG
€539.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€539.00


Estimated to ship on30 April 2025


recombinant

expressed in insect cells (S. frugiperda)

Quality Level

Assay

≥95% (SDS-PAGE)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

<1.0 EU/μg protein Endotoxin

shipped in

ambient

storage temp.

−20°C

General description

Dkk-2, Mouse, Recombinant, consists of amino acids 26-259 and expressed in S. frugiperda 21 cells with a C-terminal His•Tag sequence.
Recombinant, mouse Dickkopf related protein 2 (Dkk-2) consisting of amino acids 26-259 and expressed in S. frugiperda 21 cells with a C-terminal His•Tag sequence. Dkk proteins are expressed in mesenchymal tissues and control epithelial transformations.

Biochem/physiol Actions

Inhibition assay: Inhibits WNT-3a-induced alkaline phosphatase production in MC3T3-E1 mouse osteoblastic cells in the presence of BMP-2.



Competetive Binding Assay:

Inhibits 50% binding of biotinylated rhDkk-1 (200 ng/ml) to immobilized rmKremen (4 µg/ml) at a concentration of 5-15 µg/ml.

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 0.2 µm filtered PBS, BSA.

Reconstitution

Following reconstitution refrigerate (4°C) for short-term storage or aliquot and freeze (-20°C) for long-term storage. Stock solutions are stable for up to 1 month at 4°C or for up to 3 months at -20°C.
Reconstitute to 100 µg/ml in sterile PBS containing at least 0.1% human or bovine serum albumin.

Other Notes

Morvan, F. et al. 2006. J. Bone Miner. Res.21, 934.
Mukhopadhyay, M. et al. 2006. Development133, 2149.
Niehrs, C. 2006. Oncogene25, 7469.
Li, X. et al. 2005. Nat. Genet.37, 945.
Bullock, C.M. et al. 2004. Mol. Pharmacol.65, 582.
Mao, B. and C. Niehrs 2003. Gene302, 179.
Brott, B. and S.Y. Sokol 2002. Mol. Cell. Biol.22, 6100.
Li, L. et al. 2002. J. Biol. Chem.277, 5977.
Mao, B. et al. 2002. Nature417, 664.
Mao, B. et al. 2001. Nature411, 321.
Wu, W. et al. 2000. Current Biol.10,1611.
Monaghan, A.P. et al. 1999. Mech. Dev.87, 45.
Krupnik, V.E. et al. 1999. Gene238, 301.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service