Skip to Content
Merck
All Photos(1)

Key Documents

368020

Sigma-Aldrich

Gramicidin A

from Bacillus brevis, ≥90% (HPLC), solid, ion channel-forming pentadecapeptide, Calbiochem®

Synonym(s):

Gramicidin A, High Purity, Bacillus brevis, gA

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€412.00

€412.00


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
25 MG
€412.00

About This Item

Empirical Formula (Hill Notation):
C99H140N20O17
CAS Number:
Molecular Weight:
1882.29
MDL number:
UNSPSC Code:
51283112
NACRES:
NA.77

€412.00


Estimated to ship on20 May 2025


Request a Bulk Order

Product Name

Gramicidin A, High Purity, Bacillus brevis, Gramicidin A, High Purity, Bacillus brevis, CAS 11029-61-1, is a naturally-occurring ion channel-forming pentadecapeptide. Causes K+/H+ exchange in mitochondria in a non-voltage dependent manner.

Quality Level

description

Merck USA index - 14, 4532

Assay

≥90% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

<5% gC isomers (HPLC)

color

white to off-white

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

[nH]1c2c(c(c1)C[C@H](NC(=O)[C@H](NC(=O)[C@@H](NC(=O)[C@H](NC(=O)[C@@H](NC(=O)[C@H](NC(=O)[C@@H](NC(=O)CNC(=O)[C@@H](NC=O)C(C)C)C)CC(C)C)C)C(C)C)C(C)C)C(C)C)C(=O)N[C@H](CC(C)C)C(=O)N[C@@H](Cc7c8c([nH]c7)cccc8)C(=O)N[C@H](CC(C)C)C(=O)N[C@@H](Cc5c6c([nH]c5)c

InChI

1S/C99H140N20O17/c1-51(2)37-73(109-86(123)59(17)107-81(122)49-105-96(133)82(55(9)10)106-50-121)89(126)108-60(18)87(124)117-84(57(13)14)98(135)119-85(58(15)16)99(136)118-83(56(11)12)97(134)116-80(44-64-48-104-72-34-26-22-30-68(64)72)95(132)112-76(40-54(7)8)92(129)115-79(43-63-47-103-71-33-25-21-29-67(63)71)94(131)111-75(39-53(5)6)91(128)114-78(42-62-46-102-70-32-24-20-28-66(62)70)93(130)110-74(38-52(3)4)90(127)113-77(88(125)100-35-36-120)41-61-45-101-69-31-23-19-27-65(61)69/h19-34,45-48,50-60,73-80,82-85,101-104,120H,35-44,49H2,1-18H3,(H,100,125)(H,105,133)(H,106,121)(H,107,122)(H,108,126)(H,109,123)(H,110,130)(H,111,131)(H,112,132)(H,113,127)(H,114,128)(H,115,129)(H,116,134)(H,117,124)(H,118,136)(H,119,135)/t59-,60-,73+,74+,75+,76+,77-,78-,79-,80-,82-,83+,84+,85-/m0/s1

InChI key

ZWCXYZRRTRDGQE-LUPIJMBPSA-N

General description

Naturally occurring, ion channel-forming pentadecapeptide antibiotic that renders membranes permeable to protons and alkali metal ions. Causes red blood cell lysis at concentrations of 0.5-1.0 µg/ml. Gramicidin A causes K+/H+ exchange in mitochondria in a non-voltage dependent manner.
Naturally-occurring ion channel-forming pentadecapeptide. The association of two gA peptides causes the formation of a monovalent cation-selective ion channel. Causes K+/H+ exchange in mitochondria in a non-voltage dependent manner. Causes red blood cell lysis at 0.5-1.0 µg/ml.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Causes the formation of a monovalent cation-selective ion channel
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Harmful (C)

Sequence

HCO-Val-Gly-Ala-D-Leu-Ala-D-Val-Val-D-Val-Trp-D-Leu-Trp-D-Leu-Trp-D-Leu-Trp-NHCH₂CH₂OH

Reconstitution

Following recosntituion, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Quigley, E.P., et al. 2000. J. Membr. Biol. 174,207.
Koeppe, R.E., et al. 1995. Biochemistry34, 9299.
Ketchem, R.R., et al. 1993. Science261, 1457.
Stankovic, C.J., et al. 1990. Anal. Biochem.184, 100.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrea Maset et al.
Proceedings of the National Academy of Sciences of the United States of America, 118(2) (2020-12-31)
Impairments of inhibitory circuits are at the basis of most, if not all, cognitive deficits. The impact of OPHN1, a gene associate with intellectual disability (ID), on inhibitory neurons remains elusive. We addressed this issue by analyzing the postnatal migration
Yushan Wan et al.
Cell reports, 33(5), 108346-108346 (2020-11-05)
Complex febrile seizures (FSs) lead to a high risk of intractable temporal lobe epilepsy during adulthood, yet the pathological process of complex FSs is largely unknown. Here, we demonstrate that activated microglia extensively associated with glutamatergic neuronal soma displace surrounding

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service