Skip to Content
Merck
All Photos(1)

Key Documents

364206

Sigma-Aldrich

GM 6001

≥95% (HPLC), liquid, Matrix metalloproteinase inhibitor, Calbiochem

Synonym(s):

InSolution GM 6001

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€130.00

€130.00


Estimated to ship on29 May 2025



Select a Size

Change View
1 MG
€130.00

About This Item

Empirical Formula (Hill Notation):
C20H28N4O4
Molecular Weight:
388.46
UNSPSC Code:
12352200
NACRES:
NA.77

€130.00


Estimated to ship on29 May 2025


Product Name

GM 6001, InSolution, ≥95%, broad-spectrum inhibitor of MMPs

Quality Level

Assay

≥95% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

shipped in

wet ice

storage temp.

2-8°C

General description

A potent broad-spectrum hydroxamic acid inhibitor of matrix metalloproteinases (MMPs). Inhibits MMPs in vitro [Ki = 0.4 nM for skin fibroblast collagenase (MMP-1); Ki = 0.5 nM for gelatinase A (MMP-2); Ki = 27 nM for stromelysin (MMP-3); Ki = 0.1 nM for neutrophil collagenase (MMP-8); and Ki = 0.2 nM for gelatinase B (MMP-9)]. Also prevents the release of TNF-α in vivo and in vitro and abrogates endotoxin-induced lethality in mice.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
MMP-1
Product does not compete with ATP.
Reversible: no
Target Ki: 400 pM for MMP-1; 500 pM for MMP-2; 27 nM for MMP-3; 100 pM for MMP-8; and 200 pM for MMP-9

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

A 10 mM (1 mg/257 µl) solution of GM6001 (Cat. No. 364205) in DMSO.

Reconstitution

Following initial use, aliquot and refrigerate (4°C).

Other Notes

Solorzano, C.C., et al. 1997. Shock7, 427.
Galardy, R.E., et al. 1994. Ann. NY Acad. Sci.732, 315.
Galardy, R.E., et al. 1994. Cancer Res.54, 4715.
Galardy, R.E., et al. 1993. Drugs Future18, 1109.
Grobelny, D., et al. 1992. Biochemistry31, 7152.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - closed cup - (Dimethylsulfoxide)

Flash Point(C)

87 °C - closed cup - (Dimethylsulfoxide)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karin Edsparr et al.
Immunology and cell biology, 87(6), 489-495 (2009-05-13)
Matrix metalloproteinases (MMPs) are thought to be of importance for the migratory ability of natural killer (NK) cells. Their expression and production may influence the amount of tumour-infiltrating NK cells and thereby any therapeutic capability. In this study, we sought
Neus Bota-Rabassedas et al.
Cell reports, 35(3), 109009-109009 (2021-04-22)
Cancer cells function as primary architects of the tumor microenvironment. However, the molecular features of cancer cells that govern stromal cell phenotypes remain unclear. Here, we show that cancer-associated fibroblast (CAF) heterogeneity is driven by lung adenocarcinoma (LUAD) cells at
Jina Yun et al.
Nature communications, 14(1), 156-156 (2023-01-12)
Cellular senescence and the senescence-associated secretory phenotype (SASP) are implicated in aging and age-related disease, and SASP-related inflammation is thought to contribute to tissue dysfunction in aging and diseased animals. However, whether and how SASP factors influence the regenerative capacity

Related Content

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service