Skip to Content
Merck
All Photos(1)

Key Documents

362300

Sigma-Aldrich

N-Glycosidase F, Chryseobacterium meningosepticum, Recombinant, E. coli

Synonym(s):

N-Glycosidase F, Chryseobacterium meningosepticum, Recombinant, E. coli, Peptide-N-glycosidase F, Peptide-N⁴-(acetyl-β-glucosaminyl)-asparagine Amidase, PNGase F, Glycopeptidase F

Sign Into View Organizational & Contract Pricing

Select a Size

1 SET
€622.00

€622.00


Estimated to ship on25 May 2025



Select a Size

Change View
1 SET
€622.00

About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.54

€622.00


Estimated to ship on25 May 2025


recombinant

expressed in E. coli

Quality Level

form

liquid

specific activity

≥10 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

foreign activity

β-N-acetylhexosaminidase, α-fucosidase, β-galactosidase, α- and β-mannosidase, proteases, none detected

shipped in

wet ice

storage temp.

−20°C

General description

Note: 1 mU = 1 milliunit.
Recombinant, Chryseobacterium meningosepticum N-Glycosidase F expressed in E. coli. This enzyme from Chryseobacterium (formerly Flavobacterium) meningosepticum catalyzes the hydrolysis of N-glycans from high mannose, hybrid and complex glycopeptides, and from glycoproteins generating asparagine-linked oligosaccharides. Supplied with a series of buffers.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will completely catalyze the release of N-linked oligosaccharides from 1 µmol denatured ribonuclease in 1 min at 37°C, pH 7.5.

Physical form

Enzyme supplied in 50 mM NaCl, 20 mM Tris-HCl, 1 mM EDTA, pH 7.5.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Fan, J.-Q., and Lee, Y.C. 1997. J. Biol. Chem. 272, 27058.
Altmann, F., et al. 1995. Glycoconj. J. 12, 84.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service