Skip to Content
Merck
All Photos(2)

Documents

361559

Sigma-Aldrich

GSK-3 Inhibitor XVI

GSK-3 Inhibitor XVI - CAS 252917-06-9, is a cell-permeable, potent, ATP-competitive, and highly selective GSK-3 inhibitor (IC₅₀ = 10 and 6.7 nM against GSK-3α and GSK-3β, respectively).

Synonym(s):

GSK-3 Inhibitor XVI, 6-(2-(4-(2,4-Dichlorophenyl)-5-(4-methyl-1H-imidazol-2-yl)-pyrimidin-2-ylamino)ethyl-amino)-nicotinonitrile, CHIR99021

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H18Cl2N8
CAS Number:
Molecular Weight:
465.34
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.28

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light beige

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C22H18Cl2N8/c1-13-10-29-21(31-13)17-12-30-22(32-20(17)16-4-3-15(23)8-18(16)24)27-7-6-26-19-5-2-14(9-25)11-28-19/h2-5,8,10-12H,6-7H2,1H3,(H,26,28)(H,29,31)(H,27,30,32)

InChI key

AQGNHMOJWBZFQQ-UHFFFAOYSA-N

General description

A cell-permeable aminopyrimidine compound that acts as potent, ATP-competitive, and highly selective GSK-3 inhibitor (IC50 = 10 and 6.7 nM against GSK-3α and GSK-3β, respectively). CHIR99021 exhibits little or no activity toward 46 other enzymes (IC50 >8.8 µM against 20 kinases and Ki ≥5 µM against 23 non-kinase enzymes) and shows only weak binding to 22 pharmacologically relevant receptors (Kd ~4 µM). CHIR99021 is orally available and is shown to effectively improve glucose metabolism in both murine and rat type 2 diabetes models in vivo.
A cell-permeable aminopyrimidine compound that acts as potent, ATP-competitive, and highly selective GSK-3 inhibitor (IC50 = 10 and 6.7 nM against GSK-3α and GSK-3β, respectively). Exhibits little or no activity toward 46 other enzymes (IC50 >8.8 µM against 20 kinases and Ki ≥5 µM against 23 non-kinase enzymes) and shows only weak binding to 22 pharmacologically relevant receptors (Kd ~4 µM). This compound is orally available and is shown to effectively improve glucose metabolism in both murine and rat type 2 diabetes models in vivo. Also available as a 25 mM solution in DMSO (Cat. No. 361571).

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Mussmann, R., et al. 2007. J. Biol. Chem.282, 12030.
Ring, D.B., et al. 2003. Diabetes52, 588.
Bennett, C.N., et al. 2002. J. Biol. Chem.277, 30998.
Cline, G.W., et al. 2002. Diabetes51, 2903.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fabian Philipp Kreutzer et al.
Basic research in cardiology, 117(1), 9-9 (2022-03-03)
Cardiac fibroblasts constitute the major cell type of the murine and human heart. Once activated, they contribute to an excessive deposition of extracellular matrix (ECM) leading to cardiac fibrosis and subsequently organ dysfunction. With the exception of the pulmonary drugs
Phillip Wulfridge et al.
eLife, 10 (2021-02-24)
R-loops are three-stranded nucleic acid structures with essential roles in many nuclear processes. However, their unchecked accumulation is associated with genome instability and is observed in neurodevelopmental diseases and cancers. Genome-wide profiling of R-loops in normal and diseased cells can
Giuliana Rossi et al.
Cell stem cell, 28(2), 230-240 (2020-11-12)
Organoids are powerful models for studying tissue development, physiology, and disease. However, current culture systems disrupt the inductive tissue-tissue interactions needed for the complex morphogenetic processes of native organogenesis. Here, we show that mouse embryonic stem cells (mESCs) can be
Stoyan G Petkov et al.
Methods in molecular biology (Clifton, N.J.), 2454, 717-729 (2021-03-19)
The generation and culture of transgene-free induced pluripotent stem cells (iPSCs) from the common marmoset (Callithrix jacchus) present unique challenges due to the fact that the protocols developed for culture of human or mouse pluripotent cells are not sufficiently optimized
W F Theeuwes et al.
Biochimica et biophysica acta. Molecular basis of disease, 1863(12), 3075-3086 (2017-09-26)
Aberrant skeletal muscle mitochondrial oxidative metabolism is a debilitating feature of chronic diseases such as chronic obstructive pulmonary disease, type 2 diabetes and chronic heart failure. Evidence in non-muscle cells suggests that glycogen synthase kinase-3β (GSK-3β) represses mitochondrial biogenesis and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service