Skip to Content
Merck
All Photos(1)

Key Documents

361311

Millipore

Glycodeoxycholic Acid, Sodium Salt

Induces apoptosis in hepatocytes by a mechanism associated with DNA cleavage by endonucleases. Aggregation number: 2.

Synonym(s):

Glycodeoxycholic Acid, Sodium Salt, GDCA

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
Inquire


Estimated to ship on18 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
Inquire

About This Item

Empirical Formula (Hill Notation):
C26H42NO5 · Na
CAS Number:
Molecular Weight:
471.61
MDL number:
UNSPSC Code:
12161902
NACRES:
NA.21


Estimated to ship on18 May 2025


Request a Bulk Order

Quality Level

description

RTECS - MB9670000

Assay

≥96% (TLC)

form

solid

mol wt

micellar wt 900

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

aggregation number

2

color

white

pKa 

4.8

CMC

2.1 mM

solubility

water: 10 mg/mL

shipped in

ambient

storage temp.

15-25°C

SMILES string

[Na].N(CC(=O)O)C(=O)CC[C@H]([C@@H]1[C@@]2([C@H]([C@H]3[C@@H]([C@@]4([C@@H](C[C@@H](CC4)O)CC3)C)C[C@@H]2O)CC1)C)C

InChI

1S/C26H43NO5.Na/c1-15(4-9-23(30)27-14-24(31)32)19-7-8-20-18-6-5-16-12-17(28)10-11-25(16,2)21(18)13-22(29)26(19,20)3;/h15-22,28-29H,4-14H2,1-3H3,(H,27,30)(H,31,32);/t15-,16-,17-,18+,19-,20+,21+,22+,25+,26-;/m1./s1

InChI key

IIYSTUUVIASHBG-YEUHZSMFSA-N

General description

Induces apoptosis in hepatocytes by a mechanism associated with DNA cleavage by endonucleases. Aggregation number: 2.
Ionic detergent with a CMC of 2.1 mM and a micellar weight of 900. Induces apoptosis in hepatocytes by a mechanism associated with DNA cleavage by endonucleases. Aggregation number: 2.

Warning

Toxicity: Irritant (B)

Other Notes

Kwo, P., et al. 1995. Am. J. Physiol.268, G613.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service