Skip to Content
Merck
All Photos(1)

Key Documents

36-012

Sigma-Aldrich

Anti-nitro-α/β-Synuclein (Tyr39) Antibody, clone nSyn14

clone nSyn14, Upstate®, from mouse

Synonym(s):

Anti-NACP, Anti-PARK4, Anti-PD1

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
€600.00

€600.00


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
200 μL
€600.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€600.00


Estimated to ship on23 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

nSyn14, monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

nitration (Tyr39)

Gene Information

human ... SNCA(6622)

Specificity

α- or β-synuclein nitrated at Tyr39

Immunogen

full-length recombinant human α-Synuclein nitrated in vitro

Application

Anti-nitro-α/β-Synuclein (Tyr39) Antibody, clone nSyn14 detects level of nitro-α/β-Synuclein (Tyr39) & has been published & validated for use in WB, IH.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases

Quality

routinely evaluated by immunoblot on recombinant protein nitrated in vitro

Target description

α-Synuclein, Mr 14.5kDa
β-Synuclein, Mr 17kDa

Physical form

Ascites
Format: Purified
ascites with 0.05% sodium azide

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hugo Vicente Miranda et al.
Scientific reports, 7(1), 13713-13713 (2017-10-24)
Parkinson's disease (PD) is a progressive neurodegenerative disorder known for the typical motor features associated. Pathologically, it is characterized by the intracellular accumulation of alpha-synuclein (aSyn) in Lewy bodies and Lewy neurites. Currently, there are no established biochemical markers for
Novel antibodies to synuclein show abundant striatal pathology in Lewy body diseases
Duda, J. E., et al
Annals of Neurology, 52, 205-210 (2002)
J M Souza et al.
The Journal of biological chemistry, 275(24), 18344-18349 (2000-04-05)
Intracellular proteinaceous aggregates are hallmarks of many common neurodegenerative disorders, and recent studies have shown that alpha-synuclein is a major component of several pathological intracellular inclusions, including Lewy bodies in Parkinson's disease (PD) and glial cell inclusions in multiple system
Emilio Fernández et al.
Antioxidants & redox signaling, 21(15), 2143-2148 (2014-08-16)
The research group has detected nitrosative stress and a singular version of nitrosylated serum α-synuclein in serum of Parkinson's disease (PD) patients. Dysfunction of the thyroid gland has been proposed to be linked to this disease. The aim of the
Pedro Magalhães et al.
NPJ Parkinson's disease, 8(1), 93-93 (2022-07-23)
Parkinson's disease (PD), the second most common progressive neurodegenerative disease, develops and progresses for 10-15 years before the clinical diagnostic symptoms of the disease are manifested. Furthermore, several aspects of PD pathology overlap with other neurodegenerative diseases (NDDs) linked to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service