Skip to Content
Merck
All Photos(2)

Key Documents

351801

Sigma-Aldrich

Glucose Transporter Inhibitor III, STF-31

The Glucose Transporter Inhibitor III, STF-31 controls the biological activity of Glucose Transporter.

Synonym(s):

Glucose Transporter Inhibitor III, STF-31, STF-31

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€150.00

€150.00


Estimated to ship on22 May 2025



Select a Size

Change View
10 MG
€150.00

About This Item

Empirical Formula (Hill Notation):
C23H25N3O3S
Molecular Weight:
423.53
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€150.00


Estimated to ship on22 May 2025


Quality Level

Assay

≥99% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL

shipped in

wet ice

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(NCc2ccc(cc2)C(=O)Nc3cnccc3)c1ccc(cc1)C(C)(C)C

InChI key

NGQPRVWTFNBUHA-UHFFFAOYSA-N

General description

A cell-permeable sulfonamide that selectively inhibits the growth of VHL-deficient renal cell carcinomas (RCCs) dose-dependently (0-5 µM) by directly targeting glucose transporter 1 (GLUT1), which is up-regulated by HIF (hypoxia-inducible factor) transcription factor in VHL-deficient cells. It elicits decreased oxidative phosphorylation associated with aerobic glycolysis and leads to necrosis, which is consistent with the effect induced by GLUT1 RNA interference. It does not bind to other glucose transporters, and does not inhibit a broad range of 50 tested kinases.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Chan, D., et al. 2011. Sci. Transl. Med.3, 94ra70.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service