Skip to Content
Merck
All Photos(1)

Key Documents

345991

Sigma-Aldrich

Rabbit γ-Globulin, Serum

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€528.00

€528.00


Estimated to ship on23 March 2025



Select a Size

Change View
5 G
€528.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.25

€528.00


Estimated to ship on23 March 2025


Assay

≥95% (electrophoresis)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

shipped in

ambient

storage temp.

2-8°C

General description

Native γ-globulin from rabbit serum.
Native γ-globulin from rabbit serum. Immunoglobulins (Igs) are glycoproteins found in serum and tissue fluids of all mammals. Some Igs act as cell surface receptors and others are antibodies that travel in the blood or lymph. Ig molecules have two functional regions, one of which binds antigens and another which controls effector functions, such as binding to host cells. Structurally, Igs have two identical heavy polypeptide chains and two identical light polypeptide chains that are linked by disulfide bonds. The heavy chains determine an Ig′s class and subclass (isotype). Most higher mammals have five different classes of immunoglobulins: IgG, IgA, IgM, IgD, and IgE. The Calbiochem® immunoglobulins are rigorously analyzed for purity and identity. Most are tested by using immunodiffusion, immunoelectrophoresis and SDS-PAGE.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Reconstitute in 135 mM NaCl in Milli-Q Water

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service