Skip to Content
Merck
All Photos(2)

Key Documents

345814-M

Sigma-Aldrich

Gentamycin Sulfate

≥590 μg/mg, powder, NMD inhibitor, Calbiochem®

Synonym(s):

Gentamycin Sulfate, NMD Inhibitor II

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€116.00

€116.00


Check Cart for Availability


Select a Size

Change View
1 G
€116.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

€116.00


Check Cart for Availability

Product Name

Gentamycin Sulfate, Broad-spectrum cell culture antibiotic that is nontoxic to viruses and mammalian cells at antibacterial and antimycoplasmal concentrations.

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

concentration

≥590 μg/mg (anhydrous)

color

white to off-white

solubility

water: 10 mg/mL

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(O)O.N([C@@H]7[C@H]([C@H](OC[C@@]7(O)C)O[C@@H]8[C@H]([C@@H]([C@H](C[C@H]8N)N)O[C@H]9O[C@@H](CC[C@H]9N)[C@H](NC)C)O)O)C.N([C@@H]4[C@H]([C@H](OC[C@@]4(O)C)O[C@@H]5[C@H]([C@@H]([C@H](C[C@H]5N)N)O[C@H]6O[C@@H](CC[C@H]6N)[C@H](N)C)O)O)C.N([C@@H]1[C@

InChI

1S/C21H43N5O7.C20H41N5O7.C19H39N5O7.H2O4S/c1-9(25-3)13-6-5-10(22)19(31-13)32-16-11(23)7-12(24)17(14(16)27)33-20-15(28)18(26-4)21(2,29)8-30-20;1-8(21)12-5-4-9(22)18(30-12)31-15-10(23)6-11(24)16(13(15)26)32-19-14(27)17(25-3)20(2,28)7-29-19;1-19(27)7-28-18(13(26)16(19)24-2)31-15-11(23)5-10(22)14(12(15)25)30-17-9(21)4-3-8(6-20)29-17;1-5(2,3)4/h9-20,25-29H,5-8,22-24H2,1-4H3;8-19,25-28H,4-7,21-24H2,1-3H3;8-18,24-27H,3-7,20-23H2,1-2H3;(H2,1,2,3,4)/t9-,10-,11+,12-,13+,14+,15-,16-,17+,18-,19-,20-,21+;8-,9-,10+,11-,12+,13+,14-,15-,16+,17-,18-,19-,20+;8-,9+,10-,11+,12-,13+,14+,15-,16+,17+,18+,19-;/m110./s1

InChI key

RDEIXVOBVLKYNT-HDZPSJEVSA-N

General description

Autoclavable. Stable over a wide pH range at 37°C for up to 15 days.
Broad-spectrum cell culture antibiotic that is non-toxic to viruses and mammalian cells at anti-bacterial and anti-mycoplasmal concentrations. Its extended stability and the slow development of bacterial resistance allow long-term virus and tissue culture studies. Composed of three closely related compounds: gentamycin C1, C1a, and C2. Reduces sphingomyelinase activity in fibroblasts. Molecular weight of C2 free base = 463.0.
M.W. 463.0 (C2, free base).

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 year at -20°C. Material is autoclavable. Stable over a wide pH range at 37°C for up to 15 days.

Other Notes

Montenez, J.P., et al. 1994. Toxicol. Lett. 73, 201.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service