Skip to Content
Merck
All Photos(1)

Key Documents

345742

Sigma-Aldrich

Ganglioside GD1b, Disialo, Human Brain

Gangliosides are highly purified sialic acid-containing glycolipids that are used as markers of various cell types and antigens.

Synonym(s):

Ganglioside GD1b, Disialo, Human Brain, II³(Neu5NAc)₂GgOse₄ Ceramide, Galβ1,3GalNAcβ1,4(Neu5Acα2,8)(Neu5Acα2,3)Galβ1-4Glcβ1,1ʹ Ceramide

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€411.00

€411.00


Estimated to ship on19 May 2025


Request a Bulk Order

Select a Size

Change View
1 MG
€411.00

About This Item

Empirical Formula (Hill Notation):
C84H148N4O39
Molecular Weight:
1838.08
UNSPSC Code:
12352211
NACRES:
NA.77

€411.00


Estimated to ship on19 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (TLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

chloroform/methanol (2:1): soluble

shipped in

ambient

storage temp.

−20°C

General description

Gangliosides are highly purified sialic acid-containing glycolipids that are used as markers of various cell types and antigens.
M.W. 1838.1 (based on stearoyl - 18:0).

Biochem/physiol Actions

Cell permeable: no
Primary Target
Used as markers of various cell types and antigens
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Irritant (B)

Preparation Note

Prepared from tissue of individuals that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

Glycosphingolipids

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service