Skip to Content
Merck
All Photos(1)

Key Documents

344845

Sigma-Aldrich

Fumagillin, Aspergillus fumigatus

Antiprotozoal agent and angiogenesis inhibitor.

Synonym(s):

Fumagillin, Aspergillus fumigatus

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
€139.00

€139.00


Estimated to ship on30 May 2025



Select a Size

Change View
500 μG
€139.00

About This Item

Empirical Formula (Hill Notation):
C26H34O7
CAS Number:
Molecular Weight:
458.54
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€139.00


Estimated to ship on30 May 2025


Quality Level

description

Merck USA index - 14, 4286

Assay

≥90% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white to off-white

solubility

methanol: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

O1[C@@](C1CC=C(C)C)([C@@H]2[C@]3(OC3)CC[C@H]([C@H]2OC)OC(=O)\C=C\C=C\C=C\C=C\C(=O)O)C

InChI

1S/C26H34O7/c1-18(2)13-14-20-25(3,33-20)24-23(30-4)19(15-16-26(24)17-31-26)32-22(29)12-10-8-6-5-7-9-11-21(27)28/h5-13,19-20,23-24H,14-17H2,1-4H3,(H,27,28)/b7-5+,8-6+,11-9+,12-10+/t19-,20?,23-,24-,25+,26+/m1/s1

InChI key

NGGMYCMLYOUNGM-IJIYPQOHSA-N

General description

An inhibitor of angiogenesis. Has selective cytostatic effects against ras transformed cells (IC50 = 10 pM). Antiprotozoal agent active against sporozoan parasites in bees and myxozoan parasites in fish.
Antiprotozoal agent and angiogenesis inhibitor. Acts as an inhibitor of methionine amino-peptidase 2.

Biochem/physiol Actions

Primary Target
Methionine amino-peptidase 2
Product does not compete with ATP.

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.

Other Notes

Zhang, P., et al. 2002. J. Biomed. Sci.9, 34.
Stepien, H., et al. 1996. J. Endocrinol.150, 99.
Abrams, J.S., et al. 1994. Cancer74, 1164.
Jenkins, D.C., et al. 1993. Br. J. Cancer68, 856.
Le Querrec, A., et al. 1993. Baillieres Clin. Hematol.6, 711.
Molnar, K. 1993. Acta Vet. Hung. 41, 51.
Kusada, M., et al. 1991. Biochem. Biophys. Res. Commun. 174, 1070.
Brackett, J.M., et al. 1988. J. Agric. Food Chem.36, 762.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service