Skip to Content
Merck
All Photos(1)

Key Documents

341668

Sigma-Aldrich

Rat Fibronectin

from rat plasma, liquid, , suitable for cell culture

Synonym(s):

Fibronectin, Rat Plasma

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
€330.00

€330.00


Estimated to ship on18 May 2025


Request a Bulk Order

Select a Size

Change View
500 μG
€330.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.75

€330.00


Estimated to ship on18 May 2025


Request a Bulk Order

Product Name

Fibronectin, Rat Plasma,

biological source

rat

Quality Level

description

Merck USA index - 14, 4075

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

technique(s)

cell attachment: suitable
cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−20°C

General description

Native fibronectin purified from pooled rat plasma. Effective agent for promoting attachment of cells to commonly-used culture substrates.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 100 mM NaCl, 20 mM sodium phosphate buffer, pH 7.3, preservative and reductant-free.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C). Be VERY careful during the thawing process. Place the vial in a 37°C water bath and leave it there until it is completely thawed. DO NOT DISTURB OR REMOVE THE VIAL AT ANY TIME DURING THE THAWING PERIOD. If the vial is disturbed or removed prior to complete thawing, the product will gel and be unusable. DO NOT VORTEX. DO NOT SHAKE AFTER THAWING. Mix very gently after thawing.

Analysis Note

Contaminants: none detected by IEP using anti-fibronectin and anti-rat serum
single major band by SDS-PAGE

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service