Skip to Content
Merck
All Photos(1)

Key Documents

330200-M

Sigma-Aldrich

Ecotin, E. coli

A potent, broad range inhibitor of serine proteases.

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€259.00

€259.00


Estimated to ship on28 May 2025



Select a Size

Change View
100 μG
€259.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.77

€259.00


Estimated to ship on28 May 2025


Quality Level

Assay

≥90% (SDS-PAGE)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

storage temp.

−70°C

General description

A potent, broad range inhibitor of serine proteases.
A potent, broad range inhibitor or serine proteases. Exhibits picomolar binding constant for the inhibition of chymotrypsin, elastase, Factor Xa, Factor XIIa, kallikrein, and trypsin. Also an effective inhibitor of collagenase and granzyme B.

Biochem/physiol Actions

Primary Target
serine proteases
Product does not compete with ATP.
Target IC50: 1 pM against serine proteases
binds trypsin in the picomolar range

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 1.0 mM HCl.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

McGrath, M.E., et al. 1995. Protein Sci. 4, 141.
Seong, I.S., et al. 1994. J. Biol. Chem. 269, 21915.
McGrath, M.E., et al. 1991. J. Biol. Chem. 266, 6620.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service