Skip to Content
Merck
All Photos(1)

Key Documents

328007

Sigma-Aldrich

ERK Inhibitor II, FR180204

ERK Inhibitor II, FR 180204, CAS 865362-74-9, is a cell-permeable, potent, ATP-competitive inhibitor of ERK1 and ERK2 (IC₅₀ = 510 nM and 330 nM; Ki = 310 nM and 140 nM, respectively).

Synonym(s):

ERK Inhibitor II, FR180204, 5-(2-Phenyl-pyrazolo[1,5-a]pyridin-3-yl)-1H-pyrazolo[3,4-c]pyridazin-3-ylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H13N7
CAS Number:
Molecular Weight:
327.34
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C18H13N7/c19-17-12-10-13(20-22-18(12)23-21-17)15-14-8-4-5-9-25(14)24-16(15)11-6-2-1-3-7-11/h1-10H,(H3,19,21,22,23)

InChI key

XVECMUKVOMUNLE-UHFFFAOYSA-N

General description

A cell-permeable pyrazolopyridazinamine that acts as a potent, ATP-competitive inhibitor of ERK1 and ERK2 (IC50 = 510 nM and 330 nM; Ki = 310 nM and 140 nM, respectively). Exhibits ~20-fold greater selectivity over p38α (IC50 = 10 µM) and shows little activity towards IKKα, MEK1, MKK4, PDGFRα, PKCα, Src, and Syk even at concentrations as high as 30 µM. Shown to inhibit TGF-β−induced AP-1 activation in Mv1Lu epithelial cells (IC50 = 3.1 µM). ERK Inhibitor II, Negative Control is also available (Cat. No. 328008). Also available as a 10 mM solution in DMSO (Cat. No. 328010).
A cell-permeable pyrazolopyridazinamine that acts as a potent, ATP-competitive inhibitor of ERK1 and ERK2 (IC50 = 510 nM and 330 nM; Ki = 310 nM and 140 nM, respectively). Exhibits ~20-fold greater selectivity over p38α (IC50 = 10 µM) and shows little activity towards IKKα, MEK1, MKK4, PDGFRα, PKCα, Src, and Syk even at concentrations as high as 30 µM. Shown to inhibit TGF-β−induced AP-1 activation in Mv1Lu epithelial cells (IC50 of 3.1 µM). ERK Inhibitor II, Negative Control is also available (Cat. No. 328008). Also available in InSolution format (Cat. No. 328010).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
ERK1 and ERK2
Product competes with ATP.
Reversible: no
Target IC50: 510 nM and 330 nM
Target Ki: 310 nM and 140 nM, against ERK1 and ERK2, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Ohori, M., et al. 2005. Biochem. Biophys. Res. Commun.336, 357.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiawen Yong et al.
International journal of molecular sciences, 23(15) (2022-08-13)
Ciliary neurotrophic factor (CNTF) was identified as a survival factor in various types of peripheral and central neurons, glia and non-neural cells. At present, there is no available data on the expression and localization of CNTF-receptors in cementoblasts as well
Jiawen Yong et al.
Journal of cellular and molecular medicine, 25(20), 9710-9723 (2021-09-16)
Hypoxia-induced apoptosis of cementoblasts (OCCM-30) may be harmful to orthodontic treatment. Hypoxia-inducible factor 1-alpha (HIF-1α) mediates the biological effects during hypoxia. Little is known about the survival mechanism capable to counteract cementoblast apoptosis. We aimed to investigate the potential roles
G Ruiz-Heiland et al.
Clinical oral investigations, 25(4), 1933-1944 (2020-08-21)
Juvenile obesity is a complex clinical condition that is present more and more frequently in the daily orthodontic practice. Over-weighted patients have an impaired bone metabolism, due in part to their increased levels of circulating adipokines. Particularly, leptin has been
Zahra S Thirouin et al.
The Journal of biological chemistry, 298(5), 101840-101840 (2022-03-22)
Posttranslational addition of a small ubiquitin-like modifier (SUMO) moiety (SUMOylation) has been implicated in pathologies such as brain ischemia, diabetic peripheral neuropathy, and neurodegeneration. However, nuclear enrichment of SUMO pathway proteins has made it difficult to ascertain how ion channels
Thomas Lanzicher et al.
Frontiers in physiology, 11, 173-173 (2020-04-08)
Titin is a large filamentous protein that forms a sarcomeric myofilament with a molecular spring region that develops force in stretched sarcomeres. The molecular spring has a complex make-up that includes the N2A element. This element largely consists of a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service