Skip to Content
Merck
All Photos(1)

Documents

324840

Sigma-Aldrich

EGFR/ErbB-2/ErbB-4 Inhibitor - CAS 881001-19-0 - Calbiochem

The EGFR/ErbB-2/ErbB-4 Inhibitor, also referenced under CAS 881001-19-0, controls the biological activity of EGFR/ErbB-2/ErbB-4. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

EGFR/ErbB-2/ErbB-4 Inhibitor - CAS 881001-19-0 - Calbiochem, N-(4-((3-Chloro-4-fluorophenyl)amino)pyrido[3,4-d]pyrimidin-6-yl)2-butynamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H11ClFN5O · 2.5H2O
CAS Number:
Molecular Weight:
400.79
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 4 mg/mL (Use fresh DMSO only.)

shipped in

ambient

storage temp.

−20°C

General description

A cell-permeable, ATP-binding site-targeting alkynamidopyrimidine compound that acts as a potent and irreversible inhibitor of erbB activities (IC50 = 0.3, 1.1, and 0.5 nM for erbB-1, erbB-2, and erbB-4, respectively). Inhibits EGF- and heregulin-induced erbB autophosphorylation in NIH3T3-erbB-1 and in MDA-MB-453 cells (IC50 = 2.5 and 24 nM, respectively).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
EGFR/ErbB-2/ErbB-4
Product does not compete with ATP.
Reversible: no
Target IC50: 0.3, 1.1, and 0.5 nM for erbB-1, erbB-2, and erbB-4, respectively; 2.5 and 24 nM against EGF- and heregulin-induced erbB autophosphorylation in NIH3T3-erbB-1 and in MDA-MB-453 cells, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Klutchko, S.R., et al. 2006. J. Med. Chem.49, 1475.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sylvester R Klutchko et al.
Journal of medicinal chemistry, 49(4), 1475-1485 (2006-02-17)
Structure-activity relationships for inhibition of erbB1, erbB2, and erbB4 were determined for a series of alkynamide analogues of quinazoline- and pyrido[3,4-d]pyrimidine-based compounds. The compounds were prepared by coupling the appropriate 6-aminoquinazolines or 6-aminopyrido[3,4-d]pyrimidines with alkynoic acids, using EDCI.HCl in pyridine.
Marie-Krystel Gauthier et al.
The European journal of neuroscience, 38(5), 2693-2715 (2013-06-14)
Spinal cord injury (SCI) results in degeneration of oligodendrocytes that leads to demyelination and axonal dysfunction. Replacement of oligodendrocytes is impaired after SCI, owing to the improper endogenous differentiation and maturation of myelinating oligodendrocytes. Here, we report that SCI-induced dysregulation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service