Skip to Content
Merck
All Photos(1)

Key Documents

317701

Sigma-Aldrich

Dkk Inhibitor II, IIIC3a

Synonym(s):

Dkk Inhibitor II, IIIC3a, 9-Carboxy-3-(dimethyliminio)-6,7-dihydroxy-10-methyl-3H-phenoxazin-10-ium iodide

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€221.00

€221.00


Estimated to ship on19 May 2025



Select a Size

Change View
25 MG
€221.00

About This Item

Empirical Formula (Hill Notation):
C16H16I2N2O5
Molecular Weight:
570.12
UNSPSC Code:
12352200
NACRES:
NA.77

€221.00


Estimated to ship on19 May 2025


Assay

≥90% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

black

solubility

DMSO: 10 mg/mL
hydrogen: 2.5 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

An enhanced in solution stable gallocyanine analog that directly and reversibly targets human LRP5/6 (low density lipoprotein-related protein 5/6), and disrupts its interaction with Dkk (Dickkopf) in a competitive manner. Shown to potently reverse DKK1/2/4-mediated inhibition of WNT3A-stimulated reporter gene activity (IC50 ~ 10 - 30 µM in in NIH 3T3 cells) with no effects on sFRP4-mediated inhibition of Wnt signaling. Blocks Dkk-2 - alkaline phosphatase binding to cells expressing LRP5 (IC50 = 1 µM in LRP5 transfected NIH 3T3 cells). Reduces blood glucose levels, improves glucose tolerance, and increases insulin sensitivity in type 2 diabetic mice (1.5 mg/kg, i.p. daily), mainly via its antagonistic action on DKK2. These effects are not observed in DKK2 -/- mice.

Biochem/physiol Actions

Primary Target
LRP5/6
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.
Use only fresh DMSO for reconstitution.

Other Notes

Li, X., et al. 2012. Proc. Natl. Acad. Sci. USA109, 11402.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service