Skip to Content
Merck
All Photos(1)

Key Documents

2710-OP

Millipore

Boric Acid

OmniPur® Grade, >= 99.5%

Synonym(s):

Boric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3BO3
CAS Number:
Molecular Weight:
61.83
MDL number:
UNSPSC Code:
12352200

Pricing and availability is not currently available.

vapor pressure

2.6 mmHg ( 20 °C)

Quality Level

Assay

≥99.5%

form

granular

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

mp

160 °C (dec.) (lit.)

solubility

water: 49.2 g/L at 20 °C

density

1.489 g/cm3 at 23 °C

Looking for similar products? Visit Product Comparison Guide

General description

Molecular Biology Grade

Specifications:
A (260 nm, 0.1 M, water): ≤0.01AU
A (280 nm, 0.1 M, water): ≤0.01AU
Assay: ≥99.5%
DNase: None detected
Heavy metals (as Pb): ≤0.001%
Phosphatase: None detected
Protease: None detected
RNase: None detected





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service