Skip to Content
Merck
All Photos(2)

Key Documents

239822

Sigma-Aldrich

CXCR4 Antagonist III

The CXCR4 Antagonist III controls the biological activity of CXCR4. This small molecule/inhibitor is primarily used for Biochemicals applications.

Synonym(s):

CXCR4 Antagonist III, N²-(4-((3-(Cyclohexylamino)propylamino)methyl)benzyl)-N⁴-(piperidin-4-yl) quinazoline-2,4-diamine, 2HCl, Fusin Antagonist III

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€255.00

€255.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€255.00

About This Item

Empirical Formula (Hill Notation):
C30H43N7 · 2HCl
CAS Number:
Molecular Weight:
574.63
UNSPSC Code:
12352200
NACRES:
NA.77

€255.00


Estimated to ship on29 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

DMSO: 25 mg/mL
water: 25 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

C1(N=C(NCC2=CC=C(CNCCCNC3CCCCC3)C=C2)N=C4NC5CCNCC5)=C4C=CC=C1

InChI

1S/C30H43N7/c1-2-7-25(8-3-1)33-18-6-17-32-21-23-11-13-24(14-12-23)22-34-30-36-28-10-5-4-9-27(28)29(37-30)35-26-15-19-31-20-16-26/h4-5,9-14,25-26,31-33H,1-3,6-8,15-22H2,(H2,34,35,36,37)

InChI key

SHIKEDQVUTVKBS-UHFFFAOYSA-N

General description

A cell-permeable quinazoline compound that acts as a potent and reversible CXCR4 antagonist (IC50 = 36.2 nM for SDF-1/CXCL12 binding to hCXCR4-transfected HEK293 membrane) with selectivity over closely related human chemokine receptors CXCR2, CCR2, CCR4 and CCR5 (IC50 >10 µM). Shown to inhibit CXCL12-induced Ca2+-mobilization and cell migration in hCXCR4-HEK293 cells (IC50 = 100.1 and 41 nM, respectively), and rapidly mobilize CXCR4+, CD34+ and CD133+ stem cells from bone marrow in C57BL/6 male mice (6 mg/kg, s.c).

Biochem/physiol Actions

Cell permeable: yes
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Wu, C.H., et al. 2012. Chem. Med. Chem.7, In press.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service