Skip to Content
Merck
All Photos(1)

Key Documents

234599

Sigma-Aldrich

Conduritol B Epoxide

≥98% (HPLC), solid, Glucocerebrosidase inhibitor, Calbiochem®

Synonym(s):

Conduritol B Epoxide, CBE, 1,2-Anhydro-myo-inositol

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€410.00

€410.00


Estimated to ship on27 May 2025


Request a Bulk Order

Select a Size

Change View
100 MG
€410.00

About This Item

Empirical Formula (Hill Notation):
C6H10O5
CAS Number:
Molecular Weight:
162.14
MDL number:
UNSPSC Code:
12352119
NACRES:
NA.77

€410.00


Estimated to ship on27 May 2025


Request a Bulk Order

Product Name

Conduritol B Epoxide, Conduritol B Epoxide, CAS 6090-95-5, is an irreversible Inhibitor of glucocerebrosidase in neurons. Also inhibits α-glucosidase activity in a variety of species.

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

white

solubility

DMSO: soluble
water: soluble

shipped in

ambient

storage temp.

−20°C

SMILES string

O1[C@@H]2[C@H]1[C@@H]([C@H]([C@@H]([C@H]2O)O)O)O

InChI

1S/C6H10O5/c7-1-2(8)4(10)6-5(11-6)3(1)9/h1-10H/t1-,2-,3+,4+,5-,6+/m0/s1

InChI key

ZHMWOVGZCINIHW-FTYOSCRSSA-N

General description

Inhibits α-glucosidase activity in mammals, snails, sweet almonds and yeast. An irreversible, potent, and specific inhibitor of glucocerebrosidase in cultured neurons. Has also been shown to inhibit α-glucosidase from yeast, and rabbit intestinal sucrase-isomaltase complex.

Biochem/physiol Actions

Cell permeable: no
Primary Target
α-glucosidase
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Harmful (C)

Reconstitution

Unstable in solution; reconstitute just prior to use.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oeystein Roed Brekk et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(44), 27646-27654 (2020-10-17)
Neurons are dependent on proper trafficking of lipids to neighboring glia for lipid exchange and disposal of potentially lipotoxic metabolites, producing distinct lipid distribution profiles among various cell types of the central nervous system. Little is known of the cellular
Kelly E Glajch et al.
Proceedings of the National Academy of Sciences of the United States of America, 118(31) (2021-07-31)
Loss-of-function mutations in acid beta-glucosidase 1 (GBA1) are among the strongest genetic risk factors for Lewy body disorders such as Parkinson's disease (PD) and Lewy body dementia (DLB). Altered lipid metabolism in PD patient-derived neurons, carrying either GBA1 or PD
Azucena Perez-Canamas et al.
Brain communications, 3(1), fcaa200-fcaa200 (2021-04-03)
TMEM106B is a transmembrane protein localized to the endo-lysosomal compartment. Genome-wide association studies have identified TMEM106B as a risk modifier of Alzheimer's disease and frontotemporal lobar degeneration, especially with progranulin haploinsufficiency. We recently demonstrated that TMEM106B loss rescues progranulin null
Electra Brunialti et al.
Journal of neuroinflammation, 18(1), 220-220 (2021-09-24)
Homozygotic mutations in the GBA gene cause Gaucher's disease; moreover, both patients and heterozygotic carriers have been associated with 20- to 30-fold increased risk of developing Parkinson's disease. In homozygosis, these mutations impair the activity of β-glucocerebrosidase, the enzyme encoded
Iva Stojkovska et al.
Neuron, 110(3), 436-451 (2021-11-19)
Neurodegenerative disorders are characterized by a collapse in proteostasis, as shown by the accumulation of insoluble protein aggregates in the brain. Proteostasis involves a balance of protein synthesis, folding, trafficking, and degradation, but how aggregates perturb these pathways is unknown.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service