Skip to Content
Merck
All Photos(1)

Documents

234138

Sigma-Aldrich

Human Collagen Type I

from human skin, powder

Synonym(s):

Collagen, Type I, Human Skin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.77

product name

Collagen, Type I, Human Skin,

Assay

≥95% (SDS-PAGE)

Quality Level

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤0.1% subdermal hair

solubility

acetic acid: 5 mg/mL (0.5M)

shipped in

ambient

storage temp.

2-8°C

General description

Collagen type 1 is a fibrillar collagen present abundantly in skin, bone, vasculature, and calcified tissues of teeth and bone. It is also seen in organs like the heart and lungs. Collagen type 1 is a heterotrimer consisting of two α1 (CIα1) and one α2 (CIα2) chains with a helical region made of repeating units of proline and hydroxyproline.

Application

Collagen Type I, Human Skin has been used as a skin protein to study the effect of age-related NADH oxidase (ar-NOX)-catalyzed oxidation on skin proteins. It has also been used as a control sample in High-resolution mass spectrometry (HRMS) analysis.

Biochem/physiol Actions

Collagen type 1 is a main component of the dermal extracellular matrix (ECM) and participates in skin elasticity. Mutations in collagen type 1 are associated with osteogenesis imperfecta and Ehlers–Danlos syndrome.

Warning

Toxicity: Standard Handling (A)

Physical form

Salt-free.

Preparation Note

Prepared from tissue of individuals that have been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

The recommended concentration is 0.01 to 0.5 M Acetic Acid (or another acidic buffer), with a maximum concentration of 5 mg/ml. The product has been tested at 0.5 M Acetic Acid and is stable for 1 month at 2-8°C. After reconstitution, aliquot and freeze at -20°C. Stock solutions remain stable for up to 6 months at this temperature.

Other Notes

Gallop, P.M., and Siefert, S. 1963. Methods Enzymol. 6, 635.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Gref et al.
Scientific reports, 10(1), 16883-16883 (2020-10-11)
Vitamin C (Vit C) benefits to human skin physiology notably by stimulating the biosynthesis of collagen. The main cutaneous collagens are types I and III, which are less synthesized with aging. Vit C is one of the main promotors of
Collagen type I as a ligand for receptor-mediated signaling
Frontiers in physics, 5, 12-12 (2017)
Xing Xu et al.
BMC biotechnology, 11, 69-69 (2011-06-28)
Collagens require the hydroxylation of proline (Pro) residues in their triple-helical domain repeating sequence Xaa-Pro-Gly to function properly as a main structural component of the extracellular matrix in animals at physiologically relevant conditions. The regioselective proline hydroxylation is catalyzed by
structure, function, and biomarkers
Biochemistry of Collagens, Laminins and Elastin (2019)
Christiaan Meadows et al.
Archives of dermatological research, 306(7), 645-652 (2014-06-08)
Age-related NADH oxidase (arNOX), a cell surface-located hydroquinone oxidase capable of superoxide generation, appears at age 30 and increases with age thereafter. The ectodomain of arNOX is shed from the cell surface into body fluids including sera and saliva where

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service