Skip to Content
Merck
All Photos(1)

Key Documents

233493

Sigma-Aldrich

Coagulation Factor α-XIIa, Human Plasma

Coagulation Factor α-XIIa, Human Plasma, is a native coagulation factor α-XIIa prepared from homogeneous Factor XII using dextran sulfate. Dextran sulfate is removed after activation.

Synonym(s):

Coagulation factor

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
€1,040.00

€1,040.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
500 μG
€1,040.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.25

€1,040.00


Estimated to ship on30 May 2025


Request a Bulk Order

Quality Level

form

liquid

specific activity

50 units/mg protein (clotting assay)

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−70°C

General description

Native coagulation factor α-XIIa from human plasma prepared from homogeneous Factor XII using dextran sulfate. Dextran sulfate is removed after activation. α-XIIa is a serine protease responsible for activation of Factor XI to Factor XIa. Factor XIIa and prekallikrein are thought to be involved in a reciprocal activation mechanism whereby Factor XIIa activates prekallikrein to kallikrein, which in turn converts Factor XII to Factor XIIa.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 150 mM NaCl, 4 mM sodium acetate buffer, pH 5.3.

Preparation Note

Prepared from plasma that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Analysis Note

Clotting activity is determined by comparison to a standard curve produced using normal pooled human plasma
homogeneous by SDS-PAGE

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service