Skip to Content
Merck
All Photos(1)

Key Documents

23-059-M

Sigma-Aldrich

TRAF6, active

Synonym(s):

TNF receptor-associated factor 6, Interleukin-1 signal transducer, RING finger protein 85

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€685.00
250 μG
€4,020.00

€685.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
10 μG
€685.00
250 μG
€4,020.00

About This Item

UNSPSC Code:
12352200
eCl@ss:
32160405
NACRES:
NA.41

€685.00


Estimated to ship on26 May 2025


Request a Bulk Order

biological source

human

Quality Level

form

liquid

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable

GenBank accession no.

General description

TRAF6 is a ubiquitin ligase essential for the activation of NF-κB and MAP kinases in multiple signalling pathways including those emanating from the tumour-necrosis-factor receptor (TNFR) and interleukin-1/Toll-like receptors (IL-1R/TLR). TRAF6 also transduces signals from receptors such as CD40 and RANK and may be essential for the formation of functional osteoclasts. Studies indicate that amplification of the TRAF6 locus is a somatic and frequent event in several human cancer types and is a commonly amplified oncogene bridging RAS and NF-κB in lung cancer. Inhibition of TRAF6 in lung cancer cells has been shown to suppress NF-κB activation, anchorage-independent growth and tumour formation.

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service