Skip to Content
Merck
All Photos(2)

Key Documents

23-056-M

Sigma-Aldrich

XIAP, active

Synonym(s):

Baculoviral IAP repeat-containing protein 4 (BIRC4), IAP-like protein, Inhibitor of apoptosis protein 3 (API3)

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€692.00

€692.00


Estimated to ship on20 May 2025



Select a Size

Change View
10 μG
€692.00

About This Item

UNSPSC Code:
12352204
eCl@ss:
32160405
NACRES:
NA.41

€692.00


Estimated to ship on20 May 2025


biological source

human

Quality Level

form

liquid

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable

NCBI accession no.

General description

X-linked inhibitor of apoptosis (XIAP) is one of a group of intracellular proteins which have been shown to act as endogenous inhibitors of caspases, the enzymatic effectors of apoptosis. XIAP possesses a C-terminal RING domain that functions as an E3 ubiquitin ligase and is able to ubiquitinate a variety of substrates thereby affecting a broad range of cellular activities beyond apoptotic suppression. There is evidence that XIAP plays a significant role in cell division, morphogenesis, heavy metal homeostasis, NF-kB activation and MAP kinase signalling. XIAP expression has been shown to be elevated in several malignancies and so is considered a promising target in anticancer therapies.

Application

Ubiquitination Cascade Comoponent: E3

Physical form

Purified using glutathione sepharose, followed by gel filtration

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service