Skip to Content
Merck
All Photos(2)

Key Documents

23-041-M

Sigma-Aldrich

c-Cbl Protein, active, 10 µg

Active, full length, recombinant human c-Cbl. N-terminal GST-tagged. For use in Enzyme Assays. Functions as an E3 ligase in ubiquitination assays.

Synonym(s):

E3 ubiquitin-protein ligase CBL, Casitas B-lineage lymphoma proto-oncogene, RING finger protein 55, Signal transduction protein CBL, CBL2, RNF55

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€676.00

€676.00


Estimated to ship on02 May 2025


Request a Bulk Order

Select a Size

Change View
10 μG
€676.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41

€676.00


Estimated to ship on02 May 2025


Request a Bulk Order

biological source

human

Quality Level

100
300

recombinant

expressed in baculovirus infected Sf21 cells

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable

NCBI accession no.

General description

Intracellular signals mediated by receptor tyrosine kinases play a pivotal role in morphogenesis, cell fate determination and pathogenesis. c-Cbl is a ubiquitin ligase (E3) that facilitates the regulation of these pathways, attenuating signalling through ubiquitination and subsequent protein degradation. c-Cbl contains several functional domains, including a tyrosine kinase-binding domain that associates with a number of protein tyrosine kinases in a phosphorylation-dependent manner, a catalytic RING-finger domain that interacts with E2s and is essential for E3 activity and a C-terminal proline rich domain which mediates interactions with SH3 containing proteins. Mutations in the gene for c-Cbl have been implicated in a number of human cancers, particularly acute myeloid leukaemia.
N-terminal cMyc and GST-tagged, recombinant human IKKγ full length, expressed by baculovirus in Sf21 insect cells.

Application

Ubiquitination Cascade Comoponent: E3

Physical form

Purified using glutathione sepharose.

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service