Skip to Content
Merck
All Photos(2)

Key Documents

23-022-M

Sigma-Aldrich

UbcH3 Protein, active, 10 µg

Active, full length, recombinant human UbcH3. N-terminal HA, 6His-tagged. For use in Enzyme Assays. Functions as an E2 enzyme in ubiquitination assays.

Synonym(s):

Ubiquitin-conjugating enzyme E2 R1, E2-CDC34, UBE2R1, CDC34, Ubiquitin-protein ligase R1

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€207.00

€207.00


Estimated to ship on14 May 2025


Request a Bulk Order

Select a Size

Change View
10 μG
€207.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41

€207.00


Estimated to ship on14 May 2025


Request a Bulk Order

biological source

human

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable

NCBI accession no.

General description

Ubiquitin-conjugating (E2) enzymes are characterized by the presence of a highly conserved ubiquitin-conjugating domain which accommodates ATP-activated ubiquitin (Ub) via a covalently linked thioester onto its active-site residue. E2 enzymes act via selective protein-protein interactions with the ubiquitin-activating E1 enzyme and ubiquitin ligase E3 enzymes and are able to differentiate effects on downstream substrates, either with a single Ub molecule or an Ub chain. While E3s are involved in substrate selection, E2s are the main determinants for selection of the lysine to construct Ub chains, which thereby directly control the cellular fate of the substrate.
UbcH3 has been shown to interact with a number of different E3 ligases, including SCFSkp2/Cks1, promoting the ubiquitination of p27 and SCFβTrCP1, promoting the ubiquitination of β-catenin and IκBα.

Application

Active, full length, recombinant human UbcH3. N-terminal HA, 6His-tagged. For use in Enzyme Assays. Functions as an E2 enzyme in ubiquitination assays.

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service