Skip to Content
Merck
All Photos(1)

Key Documents

219457

Sigma-Aldrich

Cdk Inhibitor, p35 - CAS 471270-60-7 - Calbiochem

Synonym(s):

Cdk Inhibitor, p35 - CAS 471270-60-7 - Calbiochem, 2-(3-Hydroxypropylamino)-6-( o-hydroxybenzylamino)-9-isopropylpurine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H24N6O2
CAS Number:
Molecular Weight:
356.42
UNSPSC Code:
12352200

Assay

≥95% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

This product has been discontinued.



An analog of Olomoucine (Cat. No. 495620) that acts as a potent inhibitor of Cdk1 (IC50 = 100 nM) and Cdk2 (IC50 = 80 nM). Also displays anti-proliferative and pro-apoptotic effects. The correlation between inhibition of Cdk1/Cdk2 and inhibition of cell proliferation suggests that the anti-proliferative effects are due, in part, to Cdk inhibition.

An analog of Olomoucine (Cat. No. 495620) that acts as a potent inhibitor of Cdk1 (IC50 = 100 nM) and Cdk2 (IC50 = 80 nM). Also displays anti-proliferative and pro-apoptotic effects.

Biochem/physiol Actions

Cell permeable: no
Primary Target
CDK1, CDK2
Product does not compete with ATP.
Reversible: no
Target IC50: 100 nM, 80 nM, against Cdk1 and Cdk2, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Vermeulen, K., et al. 2002. Leukemia16, 299.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service