Skip to Content
Merck
All Photos(1)

Documents

219415

Sigma-Aldrich

Cathepsin Inhibitor I

The Cathepsin Inhibitor I controls the biological activity of Cathepsin. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

Cathepsin Inhibitor I, Z-FG-NHO-Bz, CATI -1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H25N3O6
Molecular Weight:
475.49
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

>90% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated
protect from light

color

white

solubility

DMSO: soluble
acetonitrile: soluble
ethanol: soluble

shipped in

ambient

storage temp.

10-30°C

General description

Cell-permeable cysteine protease inhibitor. Inhibits cathepsin B (k2/Ki = 8.9 x 103 M-1 sec-1), cathepsin L (k2/Ki = 3.8 x 105 M-1 sec-1), cathepsin S (k2/Ki = 4.2 x 104 M-1 sec-1), and papain (k2/Ki = 2.4 x 103 M-1 sec-1).
Cysteine protease inhibitor. Selectively inhibits cathepsin B (k2/Ki = 8.9 x 103 M-1sec-1), cathepsin L (k2/Ki = 3.8 x 105 M-1sec-1), cathepsin S (k2/Ki = 4.2 x 104 M-1sec-1), and papain (k2/Ki = 2.4 x 103 M-1sec-1). Half-life in buffered solution (pH 5-9) = 25 h (30°C); 300 h (0°C).
Half-life in buffered solution (pH 5-9) = 25 h (30°C); 300 h (0°C).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
cathepsin B
Product does not compete with ATP.
Reversible: no
k₂/Ki = 8.9 x 10³ M⁻¹sec⁻¹ for cathepsin B

Warning

Toxicity: Standard Handling (A)

Sequence

Z-Phe-Gly-NHO-Bz

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Zhu, D. and Uckun, F.M. 2000. Clin. Cancer Res.6, 2064.
Demuth, H.-U., et al. 1996. Biochim. Biophys. Acta 1295, 179.
Brömme, D., and Demuth, H.-U. 1994. Methods Enzymol. 244, 671.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huilin Cheng et al.
Poultry science, 101(12), 102185-102185 (2022-10-12)
The proteolysis trends and meat quality of the chicken pectoralis major (PM) and iliotibialis (IL) muscles stored at 4°C for 7 d were investigated. After 7 d of storage, the purge loss was higher (P < 0.05) in PM than

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service