Skip to Content
Merck
All Photos(1)

Key Documents

219364

Sigma-Aldrich

Cathepsin B, Human Liver

Cathepsin B, Human Liver, CAS 9047-22-7, is a purified native cathepsin B from human liver, purified by affinity chromatography. Upregulated in many types of tumors.

Synonym(s):

Cathepsin B, Human Liver

Sign Into View Organizational & Contract Pricing

Select a Size

5 U
€354.00

€354.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
5 U
€354.00

About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

€354.00


Estimated to ship on25 May 2025


Request a Bulk Order

biological source

human liver

Quality Level

description

Merck USA index - 14, 1905

Assay

>95% (SDS-PAGE)

form

liquid

specific activity

≥50 units/mg protein

purified by

affinity chromatography

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

technique(s)

activity assay: suitable

pH range

3.5—6.0

suitability

suitable for molecular biology

application(s)

life science and biopharma

shipped in

wet ice

storage temp.

−70°C

Gene Information

human ... CTSB(1508)

General description

Research area: Cell Signaling

Cathepsin B, Human Liver, is a native cathepsin B from human liver, purified by affinity chromatography. Cathepsin B is a lysosomal protease with endo/exopeptidase[1] and a carboxydipeptidase activity. It is also known as cysteine cathepsin and belongs to the C1 family of papain-like enzymes. Cathepsin B like H, L, C, X, F, O and V is ubiquitously expressed in human tissues.[2]

Optimal pH: 3.5-6.0.

Application

Cathepsin B, Human Liver has been used to assess its linker cleavage or proteolytic activity using an LC/MS MRM-based quantitation method.[3][4]

Biochem/physiol Actions

Cathepsin B facilitates non-specific, bulk proteolysis within the acidic conditions of the endosomal/lysosomal compartment. Here, they break down both intracellular and extracellular proteins.[2] Upregulation and secretion of cathepsin B have been shown to occur in many types of tumors and to correlate positively with their invasive and metastatic capabilities. Cathepsin B is shown to facilitate tumor invasion by dissolving extracellular barriers.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will hydrolyze 1.0 µmol Z-RR-βNA per min at 40°C, pH 6.0 in the presence of DTT.

Physical form

In 50 mM sodium acetate buffer, 1 mM EDTA, pH 5.0.

Preparation Note

Prepared from tissue of individuals that have been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Kostoulas, G., et al. 1999. FEBS Lett. 455, 286.
Berquim, I.M., and Sloane, B.F. 1996. Adv. Exp. Med. Biol.389, 281.
Keppler, D., et al. 1994. Biochim. Biophys. Acta.1226, 117.
Musil, D., et al. 1991. EMBO J.10, 2321.
Sloane B.F. 1990. Semin. Cancer Biol.1, 137.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

BinQing Wei et al.
Journal of medicinal chemistry, 61(3), 989-1000 (2017-12-12)
Antibody-drug conjugates (ADCs) have become an important therapeutic modality for oncology, with three approved by the FDA and over 60 others in clinical trials. Despite the progress, improvements in ADC therapeutic index are desired. Peptide-based ADC linkers that are cleaved
Lysosomal pathways to cell death and their therapeutic applications
Cesen MH, et al.
Experimental Cell Research, 1245-1251 (2012)
Lorna Kategaya et al.
Nature, 550(7677), 534-538 (2017-10-19)
The ubiquitin system regulates essential cellular processes in eukaryotes. Ubiquitin is ligated to substrate proteins as monomers or chains and the topology of ubiquitin modifications regulates substrate interactions with specific proteins. Thus ubiquitination directs a variety of substrate fates including
Vito Turk et al.
Biochimica et biophysica acta, 1824(1), 68-88 (2011-10-26)
It is more than 50 years since the lysosome was discovered. Since then its hydrolytic machinery, including proteases and other hydrolases, has been fairly well identified and characterized. Among these are the cysteine cathepsins, members of the family of papain-like

Questions

  1. We have bought this product (Cathepsin B, Human Liver, 219364), and the size is 5U. We are facing a problem with the working stock concentration. Do we need to resubstitute the volume into MQ water or buffer?

    1 answer
    1. This product is prepared in 50 mM sodium acetate buffer containing 1 mM EDTA, pH 5.0. This buffer may also be used to dilute the material further. This item has not been tested with other buffers as a diluent, however 25 mM MES has also been cited in the literature. See the link below to review this reference:
      https://www.sigmaaldrich.com/US/en/technical-documents/protocol/protein-biology/enzyme-activity-assays/enzymatic-assay-of-cathepsin-b

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service