Skip to Content
Merck
All Photos(1)

Key Documents

217713

Sigma-Aldrich

Cdk1/2 Inhibitor II, NU6102

The Cdk1/2 Inhibitor II, NU6102, also referenced under CAS 444722-95-6, controls the biological activity of Cdk1/2. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Cdk1/2 Inhibitor II, NU6102, 6-Cyclohexylmethoxy-2-(4ʹ-sulfamoylanilino)purine

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€98.60

€98.60


Estimated to ship on10 May 2025



Select a Size

Change View
1 MG
€98.60

About This Item

Empirical Formula (Hill Notation):
C18H22N6O3S
CAS Number:
Molecular Weight:
402.47
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€98.60


Estimated to ship on10 May 2025


Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 20 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(N)c1ccc(cc1)Nc2nc3[nH]cnc3c(n2)OCC4CCCCC4

InChI

1S/C18H22N6O3S/c19-28(25,26)14-8-6-13(7-9-14)22-18-23-16-15(20-11-21-16)17(24-18)27-10-12-4-2-1-3-5-12/h6-9,11-12H,1-5,10H2,(H2,19,25,26)(H2,20,21,22,23,24)

InChI key

OWXORKPNCHJYOF-UHFFFAOYSA-N

General description

A cell-permeable 2,6-disubstituted purine compound that displays antiproliferative properties. Acts as a potent and ATP-competitive inhibitor of Cdk1/cyclin B and Cdk2/cyclin A3 (IC50 = 9.5 nM and 5.4 nM). Displays greater selectivity for Cdk1/2 over other kinases tested (IC50 = 600 nM, 800 nM, 900 nM and 1.6 µM for ROCKII, PDK1, DYRK1A and Cdk4/D1, respectively). Shown to inhibit human MCF-7 breast carcinoma cell growth with a GI50 of 8 µM.
A cell-permeable purine analog that displays anti-proliferative properties and acts as a potent and ATP-competitive inhibitor of Cdk1/cyclin B and Cdk2/cyclin A3 (IC50 = 9.5 nM and 5.4 nM, respectively). Displays greater selectivity for Cdk′s over other kinases tested (IC50 = 30 nM, 600 nM, 800 nM, 900 nM, and 1.6 µM for Cdk2/A, ROCKII, PDK1, DYRK1A, and Cdk4/D1, respectively). Shown to inhibit human MCF-7 breast carcinoma cell growth (GI50 of 8 µM).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Cdk1/cyclin B, Cdk2/cyclin A3
Product competes with ATP.
Reversible: no
Target IC50: 9.5 nM, 5.4 nM against Cdk1/cyclin B, Cdk2/cyclin A3, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Hardcastle, I.R., et al. 2004. J. Med. Chem.47, 3710.
Sayle, K.L., et al. 2003. Bioorg. Med. Chem. Lett.13, 3079.
Davies, T.G., et al. 2002. Nat. Struct. Biol.9, 745.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service