Skip to Content
Merck

Skip To

217538

β-Carotene

Vitamin A precursor.

Synonym(s):

β-Carotene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 g
Check Cart for Availability
Inquire

About This Item

Empirical Formula (Hill Notation):
C40H56
CAS Number:
Molecular Weight:
536.87
UNSPSC Code:
12352205
NACRES:
NA.25
MDL number:

Inquire

Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

Merck USA index - 14, 1853

Quality Segment

assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

dark red

solubility

DMSO: soluble, cyclohexane: soluble

shipped in

ambient

storage temp.

−20°C

SMILES string

C1CCC(=C(C1(C)C)\C=C\C(=C\C=C\C(=C\C=C\C=C(/C)\C=C\C=C(/C)\C=C\C2=C(CCCC2(C)C)C)\C)\C)C

InChI

1S/C40H56/c1-31(19-13-21-33(3)25-27-37-35(5)23-15-29-39(37,7)8)17-11-12-18-32(2)20-14-22-34(4)26-28-38-36(6)24-16-30-40(38,9)10/h11-14,17-22,25-28H,15-16,23-24,29-30H2,1-10H3/b12-11+,19-13+,20-14+,27-25+,28-26+,31-17+,32-18+,33-21+,34-22+

InChI key

OENHQHLEOONYIE-JLTXGRSLSA-N

General description

Vitamin A precursor.

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Environmental Hazard (N)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR1239C97501065480
assay

≥97% (HPLC)

assay

-

assay

≥93% (UV)

assay

-

form

solid

form

solid

form

powder

form

-

solubility

DMSO: soluble, cyclohexane: soluble

solubility

-

solubility

hexane: 1.1 mg/mL, DMSO: 30 μg/mL

solubility

-

storage temp.

−20°C

storage temp.

-10 to -25°C

storage temp.

−20°C

storage temp.

−20°C

color

dark red

color

-

color

dark red to brown

color

-

Quality Level

100

Quality Level

300

Quality Level

200

Quality Level

-


Still not finding the right product?

Explore all of our products under β-Carotene


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

217.9 °F - closed cup

flash_point_c

103.3 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters