Skip to Content
Merck
All Photos(1)

Key Documents

210150

Sigma-Aldrich

Cantharidic Acid

A potent inhibitor of protein phosphatase 2A (IC₅₀ = 50 nM).

Synonym(s):

Cantharidic Acid, 5ʹ-3ʹ2,3-Dicarboxy-2,3-dimethyl-1,4-epoxycyclohexane, exo-1,6-Dicarboxy- endo-1,6-dimethyl-7-oxabicylco[2,2,1]heptane

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€111.00

€111.00


Estimated to ship on29 May 2025



Select a Size

Change View
10 MG
€111.00

About This Item

Empirical Formula (Hill Notation):
C10H14O5
CAS Number:
Molecular Weight:
214.22
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.77

€111.00


Estimated to ship on29 May 2025


Quality Level

Assay

>95% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

warm ethanol: 10 mg/mL
DMSO: 25 mg/mL

shipped in

ambient

storage temp.

10-30°C

SMILES string

O1[C@@H]2[C@@]([C@@]([C@H]1CC2)(C)C(=O)O)(C)C(=O)O

InChI

1S/C10H14O5/c1-9(7(11)12)5-3-4-6(15-5)10(9,2)8(13)14/h5-6H,3-4H2,1-2H3,(H,11,12)(H,13,14)/t5-,6+,9-,10+

InChI key

NMTNUQBORQILRK-YUMGAWCOSA-N

General description

A potent inhibitor of protein phosphatase 2A (IC50 = 50 nM).
Inhibits protein phosphatase 2A (IC50 = 50 nM).

Biochem/physiol Actions

Cell permeable: no
Primary Target
PP2A
Product does not compete with ATP.
Reversible: no
Target IC50: 50 nM against protein phosphatase 2A

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliqot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Zhang, Z., et al. 1996. Biochemistry35, 1606.
Li, Y.-M., and Casida, J.E. 1992. Proc. Natl. Acad. Sci. USA89, 11867.
Graziano, M.J., et al. 1988. Mol. Pharmacol.33, 706.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service