Skip to Content
Merck
All Photos(1)

Key Documents

208880

Sigma-Aldrich

Anti-Calnexin, C-Terminal (575-593) Rabbit pAb

liquid, Calbiochem®

Synonym(s):

Anti-IP90, Anti-p88

Sign Into View Organizational & Contract Pricing

Select a Size

50 μL
€557.00

€557.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
50 μL
€557.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€557.00


Estimated to ship on29 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human, rabbit, mouse, rat, canine, guinea pig, bovine, hamster, monkey, sheep

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Anti-Calnexin, C-Terminal (575-593), rabbit polyclonal, recognizes the ~90 kDa calnexin protein. It is validated for Western blotting.
Protein A purified rabbit polyclonal antibody. Recognizes the ~90 kDa calnexin protein.
Recognizes the ~90 kDa calnexin protein.

Immunogen

Canine
a synthetic peptide (AEEDEILNRSPRNRKPRRE) corresponding to amino acids 575-593 of canine calnexin, conjugated to KLH

Application

Immunoblotting (1 µg/ml)

Warning

Toxicity: Standard Handling (A)

Physical form

In PBS, 50% glycerol, pH 7.2.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
Heat-shocked HeLa cells

Other Notes

Hammond, C., et al. 1994. J. Cell Biol. 126, 41.
Ou, W.-J., et al. 1993. Nature 364, 771.
David, V., et al. 1993. J. Biol. Chem.268, 9585.
Ahluwalia, N., et al. 1992. J. Biol. Chem.267, 10914.
Galvin, K., et al. 1992. Proc. Natl. Acad. Sci. USA89, 8452.
Hochstenbach, F., et al. 1992. Proc. Natl. Acad. Sci. USA89, 4734.
Wada, I., et al. 1991. J. Biol. Chem. 266, 19599.
Variables associated with assay conditions will dictate the optimal working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anita L Pinner et al.
Schizophrenia research, 177(1-3), 78-87 (2016-02-16)
Recent reports suggest abnormalities of neurotransmitter receptor trafficking, targeting, dendritic localization, recycling, and degradation in the brain in schizophrenia. We hypothesized that a potential explanation for these findings may be abnormal posttranslational modifications that influence intracellular targeting and trafficking of
Claudia M Wunderlich et al.
Journal of immunology (Baltimore, Md. : 1950), 188(9), 4141-4144 (2012-04-03)
Circulating IL-6 levels correlate with the severity of blood-stage malaria in humans and mouse models, but the impact of IL-6 classic signaling through membrane IL-6Rα, as well as IL-6 trans-signaling through soluble IL-6Rα, on the outcome of malaria has remained
Martin Whitham et al.
The Journal of biological chemistry, 287(14), 10771-10779 (2012-02-22)
Exercise increases the expression of the prototypical myokine IL-6, but the precise mechanism by which this occurs has yet to be identified. To mimic exercise conditions, C2C12 myotubes were mechanically stimulated via electrical pulse stimulation (EPS). We compared the responses
Shami Kanekar et al.
Journal of neuroscience research, 87(6), 1424-1434 (2008-12-31)
One premise regarding the mechanism of injury-evoked neuroregeneration is that injured cells induce the release of neurotrophic factors to trigger neurogenesis. Extracellular purine nucleotides exert multiple neurotrophic actions in the central nervous system mediated via activation of purinergic receptors. However
Carol Ann Remme et al.
Circulation, 114(24), 2584-2594 (2006-12-06)
Patients carrying the cardiac sodium channel (SCN5A) mutation 1795insD show sudden nocturnal death and signs of multiple arrhythmia syndromes including bradycardia, conduction delay, QT prolongation, and right precordial ST-elevation. We investigated the electrophysiological characteristics of a transgenic model of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service