Skip to Content
Merck
All Photos(1)

Key Documents

207005

Sigma-Aldrich

Calcineurin Assay Kit, Colorimetric

Synonym(s):

Colorimetric, PP2B Assay Kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€839.00

€839.00


Estimated to ship on01 May 2025



Select a Size

Change View
1 KIT
€839.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

€839.00


Estimated to ship on01 May 2025


usage

sufficient for 96 tests

Quality Level

manufacturer/tradename

Calbiochem®

packaging

pkg of  96-well plate(s)

storage condition

OK to freeze

detection method

colorimetric

shipped in

dry ice

storage temp.

−70°C

General description

A colorimetric assay kit for measuring calcineurin (PP2B) activity and for inhibitor screening. The assay is performed using RII Phosphopeptide Substrate (Cat. No. 207008).

Packaging

1 kit in Glass bottle

Components

Human Recombinant Calcineurin, Bovine Brain Calmodulin, RII Phosphopeptide Substrate, Assay Buffer, Phosphate Detection Reagent, Phosphate Calibration Standard, ½ volume 96-Well Plate, and a user protocol.

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS

Specifications

Assay time: 3.5 h

Storage and Stability

Store all components except the 96-Well Plate (room temperature) at -70°C for the highest stability. In particular, calcineurin must be handled with utmost care in order to retain its maximal enzymatic activity. Thaw it quickly in a water bath, maintained at room temperature, or by rubbing between fingers, then immediately store on an ice bath. Any remaining unused enzyme should be quickly refrozen by placing at -70°C. To minimize the number of freeze/thaw cycles, we recommend to aliquot the calcineurin into separate tubes and then store at -70°C.

Other Notes

Mondragon, A. et al. 1997. Biochemistry 36, 4934.
Donella-Deana, A., et al. 1994. Eur. J. Biochem. 219, 109.
Enz, A., et al. 1994. Anal. Biochem. 216, 147.
Harder, K.W., et al. 1994. Biochem. J. 298, 395.
Martin, B., et al. 1985. J. Biol. Chem. 260, 14932.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service