Skip to Content
Merck
All Photos(1)

Key Documents

205805

Millipore

Carbenicillin, Disodium Salt

Synthetic penicillin derivative for ampicillin-resistant plasmids. Blocks bacterial cell wall formation. Effective against Gram-negative bacteria.

Synonym(s):

Carbenicillin, Disodium Salt, 6-[(Carboxyphenylacetyl)-amino]-3,3-dimethyl-7-oxo-4-thia-1-aza-bicyclo[3.2.0]heptane-2-carboxylic Acid, 2Na

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H16N2O6S · 2Na
CAS Number:
Molecular Weight:
422.36
MDL number:
UNSPSC Code:
51282413

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

light beige

solubility

water: 10 mg/mL

shipped in

ambient

storage temp.

15-25°C

SMILES string

[Na+].[Na+].S1[C@H]2N([C@H](C1(C)C)C(=O)[O-])C(=O)[C@H]2NC(=O)C(c3ccccc3)C(=O)[O-]

InChI

1S/C17H18N2O6S.2Na/c1-17(2)11(16(24)25)19-13(21)10(14(19)26-17)18-12(20)9(15(22)23)8-6-4-3-5-7-8;;/h3-7,9-11,14H,1-2H3,(H,18,20)(H,22,23)(H,24,25);;/q;2*+1/p-2/t9?,10-,11+,14-;;/m1../s1

InChI key

RTYJTGSCYUUYAL-YCAHSCEMSA-L

General description

Synthetic penicillin derivative for ampicillin-resistant plasmids. Blocks bacterial cell wall formation. Effective against Gram-negative bacteria.
Synthetic penicillin derivative used for gene technology when carbenicillin-resistant plasmids are studied. Typically used at 30-50 µg/ml for selection of carbenicillin-resistant Escherichia coli.

Biochem/physiol Actions

Potency: ≥770 µg/mg

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C) for long term storage or refrigerate (4°C) for short-term storage. Aqueous stock solutions are stable for 72 h at 4°C or for 4 months at -20°C.

Other Notes

Payne, D.J., and Amyes, S.G. 1993. J. Med. Microbiol. 38, 114.
DeSutter, K., et al. 1992. Mol. Microbiol.6, 2201.
Monraie, D., et al. 1992. FEBS Lett.306, 108.
Sakurai, Y., et al. 1991. J. Bacteriol.173, 7038.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service