Skip to Content
Merck
All Photos(1)

Key Documents

205548

Sigma-Aldrich

Caffeine

Trimethylated xanthine that acts as a potent central nervous system stimulant.

Synonym(s):

Caffeine, 3,7-Dihydro-1,3,7-trimethyl-1H-purine-2,6-dione

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€135.00

€135.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
500 G
€135.00

About This Item

Empirical Formula (Hill Notation):
C8H10N4O2
CAS Number:
Molecular Weight:
194.19
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€135.00


Estimated to ship on31 May 2025


Request a Bulk Order

description

Merck USA index - 14, 1636

Quality Level

Assay

≥99% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

chloroform: 50 mg/mL

shipped in

ambient

storage temp.

10-30°C

SMILES string

[n]1(c2c(nc1)N(C(=O)N(C2=O)C)C)C

InChI

1S/C8H10N4O2/c1-10-4-9-6-5(10)7(13)12(3)8(14)11(6)2/h4H,1-3H3

InChI key

RYYVLZVUVIJVGH-UHFFFAOYSA-N

General description

Trimethylated xanthine that acts as a potent central nervous system stimulant. At higher doses, produces tachycardia and interferes with the uptake and storage of Ca2+ by the sarcoplasmic reticulum in skeletal muscle. Prevents apoptosis and cell cycle effects induced by Camptothecin (Cat. No. 208925) and topotecan in HL-60 cells.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Potent central nervous system stimulant
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Zhen, W., and Vaugha, A. T. 1995. Radiat. Res.141, 170.
Howell, L. L. 1993. Cancer Res.54, 4993.
Traganos, F., et al. 1993. Cancer Res.53, 4613.
Zahradnik, I., and Palade, P. 1993. Pflugers Arch. 424, 129.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kurtis Stefan et al.
BMC genomics, 24(1), 253-253 (2023-05-12)
Cis-regulatory elements (CRE) are critical for coordinating gene expression programs that dictate cell-specific differentiation and homeostasis. Recently developed self-transcribing active regulatory region sequencing (STARR-Seq) has allowed for genome-wide annotation of functional CREs. Despite this, STARR-Seq assays are only employed in
Han Hu et al.
Cardiovascular research, 116(5), 944-955 (2019-08-03)
The elevated expression of phospholamban (PLB) has been observed in heart failure and cardiac remodelling, inhibiting the affinity of Ca2+ pump to Ca2+ thereby impairing heart relaxation. However, the mechanisms underlying the regulation of PLB remains to be further studied.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service