Skip to Content
Merck
All Photos(1)

Key Documents

203820

Sigma-Aldrich

Guanosine 3′,5′-cyclic Monophosphate, 8-Bromo-, Sodium Salt

Cell-permeable cGMP analog that is more resistant to phosphodiesterases than cGMP.

Synonym(s):

Guanosine 3′,5′-cyclic Monophosphate, 8-Bromo-, Sodium Salt, 8-Bromo-cGMP, Br-cGMP, Na

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€214.00

€214.00


Estimated to ship on28 May 2025



Select a Size

Change View
25 MG
€214.00

About This Item

Empirical Formula (Hill Notation):
C10H10BrN5O7P · Na
CAS Number:
Molecular Weight:
446.08
MDL number:
UNSPSC Code:
41106305
NACRES:
NA.77

€214.00


Estimated to ship on28 May 2025


Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

white

solubility

water: 200 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

[P]1(=O)(O[C@@H]2[C@H](O[C@H]([C@@H]2O)[n]3c4[nH]c(n[c](c4nc3Br)=O)N)CO1)O

InChI

1S/C10H11BrN5O7P/c11-9-13-3-6(14-10(12)15-7(3)18)16(9)8-4(17)5-2(22-8)1-21-24(19,20)23-5/h2,4-5,8,17H,1H2,(H,19,20)(H3,12,14,15,18)/t2-,4-,5-,8-/m1/s1

InChI key

YUFCOOWNNHGGOD-UMMCILCDSA-N

General description

Cell-permeable cGMP analog that is more resistant to phosphodiesterases than cGMP. Preferentially activates protein kinase G. Inhibits thrombin-stimulated arachidonic acid release in human platelets.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
protein kinase G
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Aqueous stock solutions are stable for up to 1 week at -20°C.

Other Notes

Kwak, B.R., and Jongsma, H.J. 1996. Mol. Cell. Biochem. 157, 93.
Perreault, T., and DeMarte, J. 1993. Am. J. Physiol. 264, H302.
Ruth, P., et al. 1991. Eur. J. Biochem.202, 1339.
Schwarzschild, M.A., and Zigmond, R.E. 1991. J. Neurochem.56, 400.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sinisa Prelic et al.
Frontiers in cellular neuroscience, 17, 1180798-1180798 (2023-06-12)
Olfaction is a crucial sensory modality in insects and is underpinned by odor-sensitive sensory neurons expressing odorant receptors that function in the dendrites as odorant-gated ion channels. Along with expression, trafficking, and receptor complexing, the regulation of odorant receptor function

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service